Skip to Content
Merck
All Photos(1)

Key Documents

16030

Sigma-Aldrich

‘Brilliant Cresyl blue′

for microscopy (Vit.), mixture of toluidine blue and waterblue

Synonym(s):

‘Cresyl blue’

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
6035110
EC Number:
UNSPSC Code:
12171500
NACRES:
NA.47

grade

for microscopy (Vit.)

Quality Level

form

powder

technique(s)

microbe id | staining: suitable

solubility

methanol: 0.01 g/10 mL, blue to very deep blue

εmax

≥1000 at 630-640 nm in 50% ethanol

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

Analysis Note

λmax. ∼635 nm, E(1%,1 cm) ∼1000-1300 (50% Ethanol)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service