Skip to Content
Merck
All Photos(4)

Key Documents

A8423

Sigma-Aldrich

Amiodarone hydrochloride

≥98% (TLC), powder, ion channel blocker

Synonym(s):

(2-Butyl-3-benzofuranyl)[4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl]methanone hydrochloride, 2-Butyl-3-benzofuranyl-4-[2-(diethylamino)ethoxy]-3,5-diiodophenyl ketone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H29I2NO3 · HCl
CAS Number:
Molecular Weight:
681.77
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

Amiodarone hydrochloride, ≥98%

Assay

≥98%

form

powder

originator

Wyeth

storage temp.

2-8°C

SMILES string

C(=O)(C=1C=2C(OC1CCCC)=CC=CC2)C3=CC(I)=C(OCCN(CC)CC)C(I)=C3.Cl

InChI

1S/C25H29I2NO3.ClH/c1-4-7-11-22-23(18-10-8-9-12-21(18)31-22)24(29)17-15-19(26)25(20(27)16-17)30-14-13-28(5-2)6-3;/h8-10,12,15-16H,4-7,11,13-14H2,1-3H3;1H

InChI key

ITPDYQOUSLNIHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Amiodarone hydrochloride has been used to study its interaction with cysteamine to stimulate autophagy in cultured biosensor cell lines. It has also been used in larval and adult organ toxicogenomic screening to analyze the transcriptional effects of pharmaceuticals.
Amiodarone hydrochloride is a non-selective ion channel blocker. Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death. Amiodarone hydrochloride has also been used in a study to determine concentrations of thyroid disrupting substances in effluents from wastewater treatment plants.

Biochem/physiol Actions

Non-selective ion channel blocker. Amiodarone induces an immediate influx of Ca2+ in Saccharomyces cerevisiae, followed by mitochondrial fragmentation and cell death.

Features and Benefits

This compound was developed by Wyeth. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Substrates

CYP2C8/9 and CYP3A family (minor pathway) inhibitor and substrate; CYP2D6, CYP3A family, CYP2D6, CYP2A6, CYP2B6 desethyl-form inhibitor

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Lact. - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Editor?s Highlight: Transgenic Zebrafish Reporter Lines as Alternative In Vivo Organ Toxicity Models
Poon K, et al.
Toxicological Sciences, 156(1), 133-148 (2017)
Na Li et al.
Journal of environmental sciences (China), 23(4), 671-675 (2011-07-29)
It is generally known that there are many endocrine disrupting compounds (EDCs) in the effluents from wastewater treatment plants (WWTPs). Most research has focused on the occurrence of estrogenic or androgenic activities, while ignoring that there are environmental chemicals disrupting
Alex Dexter et al.
Analytical and bioanalytical chemistry, 411(30), 8023-8032 (2019-11-30)
Within drug development and pre-clinical trials, a common, significant and poorly understood event is the development of drug-induced lipidosis in tissues and cells. In this manuscript, we describe a mass spectrometry imaging strategy, involving repeated analysis of tissue sections by
P Simonen et al.
Journal of internal medicine, 288(5), 560-569 (2020-05-18)
We have earlier reported that amiodarone, a potent and commonly used antiarrhythmic drug increases serum desmosterol, the last precursor of cholesterol, in 20 cardiac patients by an unknown mechanism. Here, we extended our study to a large number of cardiac
Aateka Patel et al.
Pharmaceutics, 11(7) (2019-07-20)
'Foamy' alveolar macrophages (FAM) observed in nonclinical toxicology studies during inhaled drug development may indicate drug-induced phospholipidosis, but can also derive from adaptive non-adverse mechanisms. Orally administered amiodarone is currently used as a model of pulmonary phospholipidosis and it was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service