Skip to Content
Merck
All Photos(2)

Key Documents

S0278

Sigma-Aldrich

(±)-Sotalol hydrochloride

≥98% (TLC), powder, β-Adrenoceptor antagonist

Synonym(s):

N-[4-[1-Hydroxy-2-(isopropylamino)ethyl]phenyl]methanesulfonamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N2O3S · HCl
CAS Number:
Molecular Weight:
308.82
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

(±)-Sotalol hydrochloride, ≥98% (TLC), powder

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

solubility

H2O: 20 mg/mL

originator

Bayer

SMILES string

Cl.CC(C)NCC(O)c1ccc(NS(C)(=O)=O)cc1

InChI

1S/C12H20N2O3S.ClH/c1-9(2)13-8-12(15)10-4-6-11(7-5-10)14-18(3,16)17;/h4-7,9,12-15H,8H2,1-3H3;1H

InChI key

VIDRYROWYFWGSY-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

(±)-Sotalol hydrochloride has been used to test its effect on the cardiac action potential (AP) in human hearts.[1] It has also been used as an β- blocker to determine its effect on free intracellular calcium (Ca2+) release and whole-cell currents in mammalian cancer cells.[2]

Biochem/physiol Actions

Potent β-adrenoceptor antagonist; class III antiarrhythmic; prolongs the action potential and increases the refractory period.
Sotalol has a potential to block non-cardioselective β-adrenergic receptors. It acts as an anti-arrhythmic agent and is used as a therapeutic for equine arrhythmias. In addition, sotalol also has an ability to stop atrial fibrillation recurrence.[3]

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Bayer. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Burashnikov et al.
Circulation. Arrhythmia and electrophysiology, 5(2), 400-408 (2012-02-11)
Several clinical trials have shown that vernakalant is effective in terminating recent onset atrial fibrillation (AF). The electrophysiological actions of vernakalant are not fully understood. Here we report the results of a blinded study comparing the in vitro canine atrial
László Drimba et al.
Journal of cardiovascular pharmacology, 60(2), 208-218 (2012-05-25)
High-fat diet and consequent metabolic syndrome (MS) can lead to elevated risk for cardiac arrhythmias. This preclinical study was to investigate if cicletanine (CIC) could produce cardioprotective effects in conscious rabbits exhibiting the main symptoms of MS. NZW rabbits that
[QTc-prolongation in acute drug-poisoning. Guidelines for management from the Swedish Poisons Information Centre].
Jonas Höjer
Lakartidningen, 110(19-20), 953-955 (2013-06-12)
Kyle A Arsenault et al.
The Cochrane database of systematic reviews, 1(1), CD003611-CD003611 (2013-02-27)
Atrial fibrillation is a common post-operative complication of cardiac surgery and is associated with an increased risk of post-operative stroke, increased length of intensive care unit and hospital stays, healthcare costs and mortality. Numerous trials have evaluated various pharmacological and
Amee Shah et al.
The American journal of cardiology, 109(11), 1614-1618 (2012-03-27)
Fetal supraventricular tachycardia (SVT) and atrial flutter (AF) can be associated with significant morbidity and mortality. Digoxin is often used as first-line therapy but can be ineffective and is poorly transferred to the fetus in the presence of fetal hydrops.

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Chromatograms

application for HPLCapplication for HPLCapplication for HPLC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service