Skip to Content
Merck
All Photos(4)

Key Documents

H9636

Sigma-Aldrich

Hypoxanthine

powder, suitable for cell culture, BioReagent

Synonym(s):

6-Hydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Beilstein:
5811
EC Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

Product Name

Hypoxanthine, powder, BioReagent, suitable for cell culture

product line

BioReagent

Quality Level

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

>300 °C (lit.)

solubility

formic acid: water (2:1): 50 mg/mL

SMILES string

O=C1NC=Nc2nc[nH]c12

InChI

1S/C5H4N4O/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)

InChI key

FDGQSTZJBFJUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hypoxanthine (6-hydroxypurine), a purine derivative is a naturally occurring compound. It is the deaminated form of adenine and a breakdown product of adenosine monophosphate (AMP).

Application

Hypoxanthine is a nutrient additive for a variety of cell culture applications involving bacterial, parasite (Plasmodium falciparum) and animal cells. Hypoxanthine is a component of selection media used in hybridoma technologies.
Hypoxanthine has been used:
  • as a supplement in HEPES (4-(2-hydroxyethyl)-1-piperazineethanesulfonic acid) -buffered Roswell park memorial institute (RPMI) to culture Plasmodium falciparum strain IT/FCR3
  • in the preparation of Dulbecco′s modified eagle′s medium (DMEM) + MXH for the selection of chimeric modified vaccinia virus ankara (MVA)
  • to generate superoxide anions
  • to prepare hybridoma selection and maintenance media.

Biochem/physiol Actions

Hypoxanthine is capable of stimulating cell death. It can also induce reactive oxygen species (ROS). It results in endothelial dysfunction via apoptosis, stimulated by oxidative stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxanthine causes endothelial dysfunction through oxidative stress-induced apoptosis
Kim YJ, et al.
Biochemical and biophysical research communications, 482(4), 821-827 (2017)
Binding of Plasmodium falciparum to CD36 can be shielded by the glycocalyx
Hempel C, et al.
Malaria Journal, 16(1), 1-11 (2017)
Biochemical Methods of Estimating the Time Since Death
Human Body Decomposition, 53-90 (2016)
Construction of FWPV Chimaeric MVA
Buttigieg KR, et al.
Bio-protocol, 5(1), 1-11 (2019)
Nanika Coetzee et al.
Scientific reports, 10(1), 2355-2355 (2020-02-13)
The epigenome of the malaria parasite, Plasmodium falciparum, is associated with regulation of various essential processes in the parasite including control of proliferation during asexual development as well as control of sexual differentiation. The unusual nature of the epigenome has

Articles

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Uric acid present in animal sera impacts serum-supplemented classical cell culture systems, influencing cell culture performance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service