Skip to Content
Merck
All Photos(5)

Key Documents

X2252

Sigma-Aldrich

Xanthine Oxidase microbial

lyophilized powder, ≥7 units/mg solid

Synonym(s):

Schardingerenzyme, XO, hypoxanthine oxidase, XOD, Xanthine:oxygen oxidoreductase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

microbial

Quality Level

form

lyophilized powder

specific activity

≥7 units/mg solid

mol wt

~160 kDa

technique(s)

inhibition assay: suitable

color

brown

optimum pH

7.5-8.0

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Escherichia coli K12 ... xdhA(947116) , xdhB(947205) , xdhC(945148)

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Cycle

Xanthine Oxidase (XO), a versatile molybdoflavoprotein, is one of the forms of xanthine oxidoreductases (XORs). XO is characterized with two flavin molecules (FAD), two molybdenum atoms, and eight iron atoms bound per enzymatic unit.

Application

Xanthine Oxidase microbial has been used:
  • to determine in vitro xanthine oxidase activity
  • as a source to generate superoxide anions during superoxide dismutase assay
  • as a component of the reagent mix in colorimetric paper-based device to detect allopurinol in traditional medicine by enzymatic reaction

Biochem/physiol Actions

Also contains proteolytic activity.
Xanthine Oxidase catalyzes the hydroxylation of hypoxanthine to xanthine and xanthine to uric acid and oxygen free radicals during purine metabolism. This enzyme is also implicated in caffeine metabolism. XO also plays a vital role in the catabolism of xenobiotics including various drugs such as thiopurine drugs, containing 6-mercaptopurine, allopurinol, and uric acid. Excess of XO activity causes hyperuricemia (higher levels of uric acid in the blood), which further leads to gout.

Physical properties

Inhibitors: Reducing agents, Hg++, Ag+,MIA
Optimum pH: 7.5 - 8.0
Optimum temperature: 65oC
pH Stability: pH 6.5 - 9.0 (25oC, 15hr)
Thermal stability: below 55oC (pH 8.0, 30min)

Unit Definition

One unit will convert 1.0 μmole of xanthine to uric acid per min at pH 7.5 at 25 °C. Approx. 50% of the activity is obtained with hypoxanthine as substrate.

Physical form

Lyophilized powder containing BSA and sodium glutamate as stabilizers

Storage and Stability

Tightly closed. Dry. Keep locked up or in an area accessible only to qualified or authorized
persons

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael C Hall
Molecular and cellular biochemistry, 292(1-2), 1-11 (2006-09-20)
The cytotoxicity of five oxysterols identified in cooked fish, 7-ketocholesterol, 7beta-hydroxycholesterol, cholesterol 5alpha,6alpha-epoxide, cholestanetriol and 4-cholesten-3-one, was investigated in primary cultures of bovine ovarian granulosa cells. Cells were exposed to the oxysterols individually and to a mixture of the same
Pál Pacher et al.
Pharmacological reviews, 58(1), 87-114 (2006-03-02)
The prototypical xanthine oxidase (XO) inhibitor allopurinol, has been the cornerstone of the clinical management of gout and conditions associated with hyperuricemia for several decades. More recent data indicate that XO also plays an important role in various forms of
Adrian I Campos et al.
Molecular cell, 74(6), 1291-1303 (2019-05-03)
Alternative to the conventional search for single-target, single-compound treatments, combination therapies can open entirely new opportunities to fight antibiotic resistance. However, combinatorial complexity prohibits experimental testing of drug combinations on a large scale, and methods to rationally design combination therapies
P J Marro et al.
Pediatric research, 41(4 Pt 1), 513-520 (1997-04-01)
The effect of allopurinol to inhibit purine metabolism via the xanthine oxidase pathway in neonates with severe, progressive hypoxemia during rescue and reperfusion with extracorporeal membrane oxygenation (ECMO) was examined. Twenty-five term infants meeting ECMO criteria were randomized in a
Giuseppe Faudone et al.
Journal of medicinal chemistry, 64(11), 7156-7178 (2021-05-22)
The purine alkaloid caffeine is the most widely consumed psychostimulant drug in the world and has multiple beneficial pharmacological activities, for example, in neurodegenerative diseases. However, despite being an extensively studied bioactive natural product, the mechanistic understanding of caffeine's pharmacological

Protocols

The appearance of uric acid is measured at 293nm by spectrophotometry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service