Skip to Content
Merck
All Photos(1)

Key Documents

A7437

Sigma-Aldrich

Acetate Kinase from Escherichia coli

lyophilized powder, ≥150 units/mg protein (biuret)

Synonym(s):

ATP:Acetate phosphotransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥150 units/mg protein (biuret)

composition

Protein, 5.0-20.0%

UniProt accession no.

foreign activity

adenosine triphosphatase and NADH oxidase ≤0.01%

storage temp.

−20°C

Gene Information

Escherichia coli K12 ... ackA(946775)

Looking for similar products? Visit Product Comparison Guide

General description

Acetate kinase belongs to acetate and sugar kinase/Hsc70/actin (ASKHA) structural superfamily. It exists as a dimer and has nucleotide binding site in the cleft between N and C-terminal domain.

Application

Acetate Kinase from Escherichia coli has been used in assessing acetate kinase activity during the fermatation process of Gluconacetobacter xylinus and in kinetic modeling and steady state studies.
Acetate kinase is used to phosphorylate acetate to acetyl phosphate. Acetate Kinase from Escherichia coli has been used as part of an ATP-regenerating system to study the kinetics of agonist-stimulated transphosphatidylation .

Biochem/physiol Actions

Acetate kinase plays an important role in glycolysis. Acetate kinase phosphorylates acetate in the presence of ATP and a divalent cation, which ultimately results in the production of acetyl-CoA . Acetate kinase is also involved in the metabolism of propanoate, pyruvate and taurine.
Involved in the metabolism of propanoate, pyruvate and taurine

Unit Definition

One unit will phosphorylate 1.0 μmole of acetate to acetyl phosphate per min at pH 7.6 at 25 °C.

Physical form

Lyophilized powder containing trehalose with small amounts of potassium phosphate, magnesium chloride, and dithiothreitol

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of residual ethanol concentration on the growth of Gluconacetobacter xylinus I 2281
Kornmann H, et al.
Applied Microbiology and Biotechnology, 62(2-3), 168-173 (2003)
The role of the C8 proton of ATP in the regulation of phosphoryl transfer within kinases and synthetases
Kenyon CP, et al.
BMC Biochemistry, 12(1), 36-36 (2011)
Urkinase: structure of acetate kinase, a member of the ASKHA superfamily of phosphotransferases
Buss KA, et al.
Journal of Bacteriology, 183(2), 680-686 (2001)
S C Olson et al.
The Journal of biological chemistry, 266(26), 17236-17242 (1991-09-15)
Receptor-linked activation of phospholipase D has been demonstrated recently in a variety of intact cell types including granulocytes, but little is known about the enzyme, its cofactor requirements, and regulation. Using [3H]alkyllysophosphatidylcholine to prelable an endogenous phosphatidylcholine substrate pool in
Kuo-Hsiang Tang et al.
The Journal of biological chemistry, 285(46), 35848-35854 (2010-07-24)
The anoxygenic green sulfur bacteria (GSBs) assimilate CO(2) autotrophically through the reductive (reverse) tricarboxylic acid (RTCA) cycle. Some organic carbon sources, such as acetate and pyruvate, can be assimilated during the phototrophic growth of the GSBs, in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service