Skip to Content
Merck
All Photos(1)

Key Documents

H8629

Sigma-Aldrich

Hexokinase and Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae)

lyophilized powder

Synonym(s):

HK/G6PD

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54

form

lyophilized powder

glucose-6-phosphate dehydrogenase activity

≥75 units/mg protein (biuret)

hexokinase activity

≥65 units/mg protein

purified by

crystallization

storage temp.

−20°C

General description

Research area: Cell signaling

Hexokinase and Glucose-6-phosphate Dehydrogenase are intracellular enzymes found distributed widely in the living cells.
Enzyme Commission numbers 2.7.1.1 + 1.1.1.49

Application

Hexokinase and Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae) has been used to evaluate the adenylate kinase (Adk), activities of the mutant Adk’s. It has also been used for the determination of glucose and ATP.

Biochem/physiol Actions

Hexokinase initiates the glycolysis pathway by catalyzing the phosphorylation of glucose to Glucose-6-phosphate in the presence of ATP. Glucose-6-phosphate dehydrogenase plays a role in the subsequent oxidation of Glucose-6-phosphate in the presence of triphosphopyridine nucleotide (TPN). Hexokinase and Glucose-6-phosphate Dehydrogenase both serve as a tool in biochemical and analytical studies for determining glucose, fructose, mannose, ATP, and creatin-kinase activity.

Packaging

Package sizes are based on hexokinase units.

Components

Mixture of hexokinase and glucose-6-phosphate dehydrogenase

Unit Definition

Hexokinase Unit Definition: One unit will phosphorylate 1.0 μmole of D-glucose per min at pH 7.6 at 25 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fetal bovine serum contains biologically available ATP
Vultaggio-Poma V, et al.
Purinergic Signaling, 20(1), 83-89 (2024)
Laura Prioretti et al.
Journal of phycology, 52(6), 1094-1102 (2016-10-25)
The sulfate facilitation hypothesis suggests that changes in ocean sulfate concentration influenced the rise to dominance of phytoplankton species of the red lineage. The mechanistic reasons for this phenomenon are not yet understood. We started to address this question by
d-Glucose: Determination with Hexokinase and Glucose-6-phosphate Dehydrogenase
Slein MW
Methods of Enzymatic Analysis, 8(12), 117-130 (1965)
Effect of pH on the stability of hexokinase and glucose 6-phosphate dehydrogenase
Souza M A, et al.
Applied Biochemistry and Biotechnology, 265-272 (2002)
Samantha L Barlow et al.
The Journal of experimental biology, 220(Pt 3), 414-424 (2017-02-06)
Atlantic cod are a commercially important species believed to be threatened by warming seas near their southern, equatorward upper thermal edge of distribution. Limitations to circulatory O

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service