Skip to Content
Merck
All Photos(3)

Documents

A6508

Sigma-Aldrich

L-Aspartic acid β-hydroxamate

>98%

Synonym(s):

AAH, HDX, LAH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O4
CAS Number:
Molecular Weight:
148.12
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Aspartic acid β-hydroxamate, serine racemase inhibitor

Quality Level

Assay

>98%

form

powder

color

white to yellow

application(s)

detection

storage temp.

−20°C

SMILES string

N[C@@H](CC(=O)NO)C(O)=O

InChI

1S/C4H8N2O4/c5-2(4(8)9)1-3(7)6-10/h2,10H,1,5H2,(H,6,7)(H,8,9)/t2-/m0/s1

InChI key

ZBYVTTSIVDYQSO-REOHCLBHSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Benhaim et al.
Science advances, 6(18), eaaz8822-eaaz8822 (2020-06-05)
The influenza virus hemagglutinin (HA) fusion protein has long been viewed as a "spring-loaded" fusion machine whereby activation at low pH initiates a rapid and irreversible cascade of conformational changes that drives the membrane fusion reaction. This mechanism has shaped
Franck Danel et al.
Current drug discovery technologies, 8(1), 66-75 (2010-11-26)
Aminoacyl-tRNA synthetases (AARSs) are a structurally heterogeneous family of enzymes present in prokaryotes, archaea and eukaryotes. They catalyze the attachment of tRNA to its corresponding amino acid via an aminoacyl adenylate intermediate. Errors in protein synthesis will occur if an
N Thomasset et al.
International journal of cancer, 49(3), 421-424 (1991-09-30)
D and L isomers of aspartic acid beta-hydroxamate (respectively DAH and LAH) were compared for their in vitro and in vivo activity against the murine leukemia L5178Y and their tolerance in vivo in DBA/2 mice. DAH and LAH displayed comparable
Elaine Symanski et al.
Environmental health : a global access science source, 19(1), 39-39 (2020-04-07)
From 2006 to 2011, the City of Houston received nearly 200 community complaints about air pollution coming from some metal recycling facilities. The investigation by the Houston Health Department (HHD) found that while operating within legal limits, emissions from facilities
Véronique Sauvé et al.
Nature structural & molecular biology, 25(7), 623-630 (2018-07-04)
Mutations in the ubiquitin ligase parkin are responsible for a familial form of Parkinson's disease. Parkin and the PINK1 kinase regulate a quality-control system for mitochondria. PINK1 phosphorylates ubiquitin on the outer membrane of damaged mitochondria, thus leading to recruitment

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service