Skip to Content
Merck
All Photos(2)

Key Documents

A3809

Sigma-Aldrich

Asparaginase from Escherichia coli

lyophilized powder, 100-300 units/mg protein (biuret)

Synonym(s):

L-Asparagine Amidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

100-300 units/mg protein (biuret)

composition

Protein, ≥60%

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Asparaginase is a bacterial enzyme and also a chemotherapeutic drug.

Application

Asparaginase from Escherichia coli has been used:
  • to compare the cytotoxic effect of L-asparaginase purified from Streptomyces brollosae NEAE-115
  • as a standard in asparaginase assay to quantify asparaginase activities in various eel tissues
  • to elicit amino acid deprivation

Biochem/physiol Actions

Asparaginase (ASNase) products are usually obtained from Escherichia coli and Erwinia chrysanthemi. These enzymes can block the synthesis of protein in tumor cells. It shows high activity in the G1 phase of the cell cycle. It is capable of causing pancreatitis in leukemia patients.
Asparaginase is used in enzymatic assays and to convert asparagine to aspartic acid. Asparaginase is used to reduce the formation of acrylamide in starchy food products. It is also used as a chemotherapy agent for acute lymphoblastic leukemia . Product A3809 is from Escherichia coli and is provided as a lyophilized powder containing sodium chloride.
Asparaginase may cause cell death in leukemic cells by converting the necessary L-asparagine to aspartic acid and ammonia . Asparaginase is allosterically regulated and is crucial for proper cell functioning .

Unit Definition

One unit will liberate 1.0 μmole of ammonia from L-asparagine per min at pH 8.6 at 37 °C.

Physical form

Lyophilized powder containing sodium chloride

Preparation Note

Chromatographically purified

Analysis Note

Protein determined by biuret.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Changes in plasma angiotensin subtypes in Japanese eel acclimated to various salinities from deionized water to double-strength seawater
Wong MKS, et al.
General and Comparative Endocrinology, 178(2), 250-258 (2012)
L-Asparaginase: discovery and development as a tumor-inhibitory agent.
J D Broome
Cancer treatment reports, 65 Suppl 4, 111-114 (1981-01-01)
Antineoplastic Drugs
Pharmacology and Therapeutics for Dentistry, 530-562 (2017)
Jie Xiong et al.
EBioMedicine, 72, 103614-103614 (2021-10-11)
Metabolic reprogramming plays an essential role on lymphoma progression. Dysregulation of glutamine metabolism is implicated in natural-killer T-cell lymphoma (NKTCL) and tumor cell response to asparaginase-based anti-metabolic treatment. To understand the metabolomic alterations and determine the potential therapeutic target of
Purification, characterization and immunogenicity assessment of glutaminase free L-asparaginase from Streptomyces brollosae NEAE-115
El-Naggar NEA, et al.
BMC Pharmacology & Toxicology, 19(1), 1-15 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service