Skip to Content
Merck
All Photos(2)

Key Documents

159034

Sigma-Aldrich

Acetohydroxamic acid

98%

Synonym(s):

AHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHOH
CAS Number:
Molecular Weight:
75.07
Beilstein:
1739019
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

88-90 °C (lit.)

functional group

amine

SMILES string

CC(NO)=O

InChI

1S/C2H5NO2/c1-2(4)3-5/h5H,1H3,(H,3,4)

InChI key

RRUDCFGSUDOHDG-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Acetohydroxamic acid is a potent inhibitor of bacterial urease activity and reduces urinary ammonia levels. 2-Acetohydroxamic acid loaded floating microspheres forms an efficient drug delivery system for the treatment of Helicobacter pylori.

Application

Acetohydroxamic acid was used:
  • to study the mechanism of complexation of iron (III) with acetohydroxamic acid
  • to study the inhibitory mechanism of lansoprazole and omeprazole on Helicobacter pyloni
  • in urease inhibition studies
  • for in situ generation of nitrosocarbonylmethane as a Diels-Alder dienophile
Used in urease inhibition studies and for in situ generation of nitrosocarbonylmethane as a Diels-Alder dienophile.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the Chemical Society. Perkin Transactions 1, 1001-1001 (1991)
K Nagata et al.
Antimicrobial agents and chemotherapy, 37(4), 769-774 (1993-04-01)
The gastric proton pump inhibitor lansoprazole, its active analog AG-2000, and omeprazole dose dependently inhibited urease activity extracted with distilled water from Helicobacter pylori cells; the 50% inhibitory concentrations were between 3.6 and 9.5 microM, which were more potent than
Mechanism of iron (III) complex formation. Activation volumes for the complexation of the iron (III) ion with thiocyanate ion and acetohydroxamic acid.
Funahashi S, et al.
Inorganic Chemistry, 22(14), 2070-2073 (1983)
Chem. Abstr., 116, 124530f-124530f (1992)
R B Umamaheshwari et al.
The Journal of pharmacy and pharmacology, 55(12), 1607-1613 (2004-01-24)
This investigation is part of our ongoing effort to develop effective drug delivery systems for the treatment of Helicobacter pylori infection using polycarbonate (PC) floating microspheres as drug carriers. In an effort to augment the anti-H. pylori effect of acetohydroxamic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service