Skip to Content
Merck
All Photos(2)

Documents

A6500

Sigma-Aldrich

Acetylcholine bromide

≥99%

Synonym(s):

ACh

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3N(Br)CH2CH2OCOCH3
CAS Number:
Molecular Weight:
226.11
Beilstein:
3572117
EC Number:
MDL number:
UNSPSC Code:
51151519
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥99%

form

powder

mp

140-143 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Br-].CC(=O)OCC[N+](C)(C)C

InChI

1S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1

InChI key

ZEHGKSPCAMLJDC-UHFFFAOYSA-M

Gene Information

human ... CHRM3(1131)

Looking for similar products? Visit Product Comparison Guide

Application

Acetylcholine is an endogenous neurotransmitter at cholinergic synapses that amplifies action potential of the sarcolemma thereby inducing muscle contractions. Acetylcholine bromide is used as an acetylcholine receptor agonist to identify, characterize and differentiate among types of cholinergic receptors. Acetylcholine bromide is used as an inhibitor to identify and characterize natural and mutated butyrylcholinesterase(s).

Biochem/physiol Actions

Endogenous neurotransmitter at cholinergic synapses; amplifies action potential of the sarcolemma thereby inducing muscle contractions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott A Hollingsworth et al.
Nature communications, 10(1), 3289-3289 (2019-07-25)
Allosteric modulators are highly desirable as drugs, particularly for G-protein-coupled receptor (GPCR) targets, because allosteric drugs can achieve selectivity between closely related receptors. The mechanisms by which allosteric modulators achieve selectivity remain elusive, however, particularly given recent structures that reveal
Fábio H Silva et al.
PloS one, 11(12), e0166291-e0166291 (2016-12-10)
Sickle cell disease patients display priapism that may progress to erectile dysfunction. However, little is known about the pathophysiological alterations of corpus cavernosum in sickle cell disease. Thus, this study aimed to evaluate the functional and molecular alterations of sympathetic
James A Angus et al.
European journal of pharmacology, 804, 111-116 (2017-03-17)
Endothelin receptor antagonists are approved for pulmonary arterial hypertension. Development of selective ET Human isolated pulmonary (i.d. 5.5mm) and human radial (i.d. 3.23mm) artery ring segments were mounted in organ baths for isometric force measurement. Single concentration-contraction curves to endothelin-1
Li-Yan Zhao et al.
Acta pharmacologica Sinica, 39(5), 875-884 (2018-03-30)
Xyloketal B (Xyl-B) is a novel marine compound isolated from mangrove fungus Xylaria sp. (No 2508). We previously showed that Xyl-B promoted endothelial NO release and protected against atherosclerosis through the Akt/eNOS pathway. Vascular NO production regulates vasoconstriction in central
Rahini Kakumanu et al.
Toxins, 11(4) (2019-04-04)
Russell's viper (Daboia russelii) venom causes a range of clinical effects in humans. Hypotension is an uncommon but severe complication of Russell's viper envenoming. The mechanism(s) responsible for this effect are unclear. In this study, we examined the cardiovascular effects

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service