Skip to Content
Merck
All Photos(5)

Key Documents

O7501

Sigma-Aldrich

Sodium oleate

≥99%

Synonym(s):

cis-9-Octadecenoic acid sodium salt, Oleic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COONa
CAS Number:
Molecular Weight:
304.44
Beilstein:
4046357
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Assay

≥99%

form

powder

mp

232-235 °C (lit.)

functional group

ester

lipid type

unsaturated FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].CCCCCCCC\C=C/CCCCCCCC([O-])=O

InChI

1S/C18H34O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;

InChI key

BCKXLBQYZLBQEK-KVVVOXFISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Oleic acid is used to investigate effects on apolipoprotein B synthesis, degradation and secretion in cultured cells.

Biochem/physiol Actions

Activates protein kinase C in hepatocytes.
Oleic acid increases hepatic secretion of apolipoprotein B100 in hepatocyte cell lines and in mice . Oleic acid inhibits apolipoprotein B100 secretion at higher physiologic doses .

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gisela Bevilacqua Rolfsen Ferreira da Silva et al.
International journal of nanomedicine, 9, 867-876 (2014-03-07)
Microemulsions (MEs) are colloidal systems that can be used for drug-delivery and drug-targeting purposes. These systems are able to incorporate drugs modifying bioavailability and stability and reducing toxic effects. The jasmonate compounds belong to a group of plant stress hormones
Yun-Li Wu et al.
Journal of virology, 90(4), 1729-1740 (2015-12-08)
Hepatitis B virus (HBV) has been implicated as a potential trigger of hepatic steatosis although molecular mechanisms involved in the pathogenesis of HBV-associated hepatic steatosis still remain elusive. Our prior work has revealed that the expression level of liver fatty
Craig van Rensburg et al.
PloS one, 14(6), e0217820-e0217820 (2019-06-07)
Drug resistant-tuberculosis is a growing burden on the South African health care budget. In response the National Department of Health implemented two important strategies in 2011; universal access to drug-sensitivity testing for rifampicin with Xpert MTB/RIF as the first-line diagnostic
Xuerui Yang et al.
Cell research, 19(4), 469-486 (2009-03-05)
The present study shows that double-stranded RNA-dependent protein kinase (PKR) regulates the protein expression level and phosphorylation of Bcl-2 and plays an anti-apoptotic role in human hepatocellular carcinoma cells (HepG2). In various types of cells, saturated free fatty acids (FFAs)
Andrea Dichlberger et al.
Journal of lipid research, 52(12), 2198-2208 (2011-10-06)
Lipid droplets, also called lipid bodies (LB) in inflammatory cells, are important cytoplasmic organelles. However, little is known about the molecular characteristics and functions of LBs in human mast cells (MC). Here, we have analyzed the genesis and components of

Articles

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Prof. Randal Lee discusses iron oxide magnetic nanospheres and nanocubes design considerations for biosensing applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service