Skip to Content
Merck
All Photos(2)

Key Documents

A4312

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)−Alkaline Phosphatase antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Rabbit Anti-Mouse IgG (whole molecule)−AP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

alkaline phosphatase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

mouse

technique(s)

direct ELISA: 1:30,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50
western blot: 1:30,000 using using β-Actin in total cell extract of HeLa cells (5-10 μg/well)

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG antibodies are the smallest antibody isotype found in blood and extracellular fluid, capable of fighting bacterial and viral infection.
Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. An immunoglobulin has two heavy chains and two light chains connected by a disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Mouse consists of five immunoglobulin classes- IgM, IgG, IgA, IgD and IgE. Mouse IgG is further divided into five classes- IgG1, IgG2a, IgG2b and IgG3.

Immunogen

Purified mouse IgG.

Application

Anti-Mouse IgG (whole molecule) Alkaline Phosphatase antibody produced in rabbit has been used in immunocytochemical localization of Aβ peptides and western blot analysis.
Anti-mouse IgG (whole molecule)-alkaline phosphatase antibody can be used in immunocytochemistry.
Western blot analysis of lysates from the placental cell line 3ASubE-P-3 and U037 cells were performed using alkaline phosphatase conjugated rabbit anti- mouse IgG as the secondary at 1:2000 to recognize the mouse mom Il-8RB. Secondary was developed using bromochloroindoyl phosphate (Sigma) pH. 9.5 for 10-15 minutes.Western blot analysis of coimmunoprecipitates from human cell or rat cell vaginal extracts were performed using alkaline phosphatase conjugated rabbit anti- mouse IgG as the secondary at 1:15000 dilution.

Biochem/physiol Actions

Immunoglobulin G (IgG) participates in hypersensitivity type II and type III reactions. IgG helps in opsonization, complement fixation and antibody dependent cell mediated cytotoxicity.

Physical form

Solution in 0.05 M Tris buffer, pH 8.0, containing 1 mM MgCl2, 10 mM glycine, 1% bovine serum albumin, 50% glycerol and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Przygrodzka et al.
Reproduction (Cambridge, England), 151(2), 135-147 (2015-11-19)
In order to characterize the transition of the corpora lutea (CL) from acquisition of luteolytic sensitivity to rescue of luteal function: i) the expression of 38 factors associated with steroids, prostanoids, and angiogenic systems and ii) concentrations of the main
Fernando M Diaz et al.
Virology, 488, 61-67 (2015-11-27)
Viral vaccines have traditionally protected against disease, but for viruses that establish latent infection, it is desirable for the vaccine to reduce infection to reduce latent infection and reactivation. While seroconversion has been used in clinical trials of herpes simplex
Cesar Andres Diaz Arias et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 48(3), 419-426 (2017-02-27)
Antibodies and antibody fragments are nowadays among the most important biotechnological products, and Pichia pastoris is one of the most important vectors to produce them as well as other recombinant proteins. The conditions to effectively cultivate a P. pastoris strain
M Marcinkiewicz et al.
Journal of neurochemistry, 75(5), 2133-2143 (2000-10-18)
To define the enzymes involved in the etiology of Alzheimer's disease, we compared in mouse and human brain the mRNA levels and cellular localization of the ubiquitous beta-amyloid precursor protein (beta-APP) with those of the putative alpha-secretases ADAM10 and ADAM17
Francesca Ferranti et al.
PloS one, 8(10), e76192-e76192 (2013-10-08)
Seminoma is one of the most common Testicular Germ Cell Tumours that originates during embryonic development due to an alteration of the local niche that in turn results in a delayed or blocked differentiation of Primordial Germ Cells. The block

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service