Skip to Content
Merck

PMSF-RO

Roche

PMSF

Phenylmethylsulfonyl fluoride

Synonym(s):

Phenylmethanesulfonyl fluoride, phenylmethylsulfonyl fluoride, α-Toluenesulfonyl fluoride, Benzylsulfonyl fluoride, PMSF, Phenylmethylsulfonyl fluoride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H7FO2S
CAS Number:
Molecular Weight:
174.19
PubChem Substance ID:
UNSPSC Code:
12352202
Beilstein/REAXYS Number:
2088311
MDL number:
NACRES:
NA.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

98% (TLC)

form

solid

mol wt

174.2

packaging

pkg of 10 g (10837091001), pkg of 25 g (11359061001)

manufacturer/tradename

Roche

concentration

17-170 μg/mL

technique(s)

western blot: suitable

pKa 

7.0

mp

92-94 °C

solubility

>10 mg/mL

shipped in

ambient

storage temp.

20-25°C

SMILES string

FS(=O)(=O)Cc1ccccc1

InChI

1S/C7H7FO2S/c8-11(9,10)6-7-4-2-1-3-5-7/h1-5H,6H2

InChI key

YBYRMVIVWMBXKQ-UHFFFAOYSA-N

General description

Soluble to >10 mg/ml in isopropanol, ethanol, methanol, or 1,2-propanediol. Unstable in aqueous solution. In 100% isopropanol, stable for at least 9 months at +25°C.

Application

PMSF (C7H7O2SF) inhibits serine proteases such as chymotrypsin, trypsin, thrombin, and the cysteine protease papain (reversible by DTT treatment). It does not inhibit metalloproteases, most cysteine proteases, or aspartic proteases.
PMSF (phenylmethylsulfonyl fluoride) has been used for the preparation of protein extracts from tissues and cells prior to Western blotting.

Biochem/physiol Actions

Inhibits serine proteases such as chymotrypsin, trypsin, and thrombin, and the cysteine protease papain (reversible by DTT treatment). Does not inhibit metalloproteases, most cysteine proteases, or aspartic proteases.
PMSF (phenylmethylsulfonyl fluoride) functions as a non-specific inhibitor of multiple proteases and other enzymes, such as palmityl coenzyme A deacylase, arylsulfatase A, acetylcholinesterase, trypsin and chymotrypsin. It is an irreversible inhibitor, and sulfonylates hydroxyl groups of the serine residues present in the active site of enzymes.

Preparation Note

Activator: PK requires Mg2+ (or Mn2+, CO2+) and K+ (or NH4+, Rb+) for full activity.
Working concentration: 17 to 170 μg/ml (0.1 to 1 mM)
Working solution: Due to the short half-life of PMSF, fresh solution must be added at every purification step. Under certain conditions, such as alkaline pH values, PMSF may have a half-life of only 35 minutes. High-salt conditions (3 M), often used to precipitate proteins, may also reverse the inhibition of PMSF. Pefabloc SC is recommended as a non-toxic, water-soluble, and water-stable alternative.
Storage conditions (working solution): 15 to 25 °C
Stock solutions of PMSF in 100% isopropanol are stable for at least 9 months at 15 to 25 °C.
Soluble to > 10 mg/ml in isopropanol, ethanol, methanol, or 1,2-propanediol. Unstable in aqueous solution. In 100% isopropanol, stable for at least 9 months at 15 to 25 °C.

Analysis Note

Purity: Chromatographically homogeneous

Other Notes

Due to a short half-life, fresh solution must be added at every purification step. Under certain conditions, such as alkaline pH values, PMSF may only have a half-life of 35 min. High-salt conditions (3 M), often used to precipitate proteins, may also reverse the inhibition of PMSF. Pefabloc SC is recommended as a non-toxic, water-soluble, and water-stable alternative.
For life science research only. Not for use in diagnostic procedures.


pictograms

CorrosionSkull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



D R Compton et al.
The Journal of pharmacology and experimental therapeutics, 283(3), 1138-1143 (1998-02-12)
Anandamide is an putative endogenous cannabinoid ligand that produces pharmacological effects similar to those of Delta9-tetrahydrocannabinol, the principle psychoactive constituent in marijuana. There is considerable evidence that the enzyme inhibitor phenylmethylsulfonyl fluoride (PMSF) is capable of altering the actions of
Hongjuan Yu et al.
PloS one, 8(11), e80815-e80815 (2013-11-16)
A kinase interacting protein 1 (AKIP1) is a molecular regulator of protein kinase A and nuclear factor kappa B signalling. Recent evidence suggests AKIP1 is increased in response to cardiac stress, modulates acute ischemic stress response, and is localized to
Guilian Tian et al.
The Journal of biological chemistry, 284(28), 18980-18993 (2009-05-09)
Clarin-1 is the protein product encoded by the gene mutated in Usher syndrome III. Although the molecular function of clarin-1 is unknown, its primary structure predicts four transmembrane domains similar to a large family of membrane proteins that include tetraspanins.



Global Trade Item Number

SKUGTIN
1135906100104061838701305
1083709100104061838683991