Skip to Content
Merck
All Photos(3)

Key Documents

A3131

Sigma-Aldrich

D-(−)-Arabinose

≥98% (GC)

Synonym(s):

Aldehydo-D-arabinose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723079
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

≥98% (GC)

form

powder

optical activity

[α]20/D -105 to -103 °, c = 4% (w/v) in water

technique(s)

gas chromatography (GC): suitable

color

white to off-white

mp

162-164 °C (lit.)

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

room temp

SMILES string

O[C@@H]1COC(O)[C@@H](O)[C@@H]1O

InChI

1S/C5H10O5/c6-2-1-10-5(9)4(8)3(2)7/h2-9H,1H2/t2-,3-,4+,5?/m1/s1

InChI key

SRBFZHDQGSBBOR-ZRMNMSDTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-(−)-Arabinose is a D-ribose analogue present in sugar polymers such as the arabinogalatans of mycobacterial cell walls and plant tissues. D-Arabinose is a substrate used to identify, differentiate and characterize arabinose isomerase(s) that have commercial value in the production of tagatose, a low calorie sweetener.

Biochem/physiol Actions

D-Arabinose is a reducing sugar. It is a pentose analog of D-ribose that is a constituent of mycobacterial cell wall arabinogalactans. It is also a substrate for D-erythroascorbic acid synthesis in yeast.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariane-Saskia Ries et al.
Nature communications, 8, 15738-15738 (2017-06-07)
Major depressive disorder (MDD) affects millions of patients; however, the pathophysiology is poorly understood. Rodent models have been developed using chronic mild stress or unavoidable punishment (learned helplessness) to induce features of depression, like general inactivity and anhedonia. Here we
Astrid Rohwedder et al.
Current biology : CB, 26(5), 661-669 (2016-02-16)
Dopaminergic neurons serve multiple functions, including reinforcement processing during associative learning [1-12]. It is thus warranted to understand which dopaminergic neurons mediate which function. We study larval Drosophila, in which only approximately 120 of a total of 10,000 neurons are
Derek Thomson et al.
Pulmonary pharmacology & therapeutics, 40, 69-79 (2016-05-26)
It is now recognized that certain polysaccharides can exhibit anti-inflammatory activity, including the glycosaminoglycan (GAG) heparin that is widely used as an anti-coagulant drug. However, it would be desirable to identify molecules that retain the anti-inflammatory actions of heparin, but
L Zhang et al.
Asian-Australasian journal of animal sciences, 27(6), 855-861 (2014-07-23)
This experiment was performed to investigate the effects of exogenous xylanase supplementation on performance, nutrient digestibility and the degradation of non-starch polysaccharides (NSP) in different sections of the gastrointestinal tract (GIT) of broilers fed wheat-based diets. A total of 120
Astrid Rohwedder et al.
Chemical senses, 37(8), 711-721 (2012-06-15)
Gustatory stimuli allow an organism not only to orient in its environment toward energy-rich food sources to maintain nutrition but also to avoid unpleasant or even poisonous substrates. For both mammals and insects, sugars-perceived as "sweet"-potentially predict nutritional benefit. Interestingly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service