Skip to Content
Merck
All Photos(1)

Key Documents

A2013

Sigma-Aldrich

D-Arabinose 5-phosphate disodium salt

≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9O8PNa2
CAS Number:
Molecular Weight:
274.07
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.79

biological source

synthetic (organic)

Assay

≥95% (HPLC)

form

powder

impurities

~1% ribose 5-phosphate

color

white to off-white

solubility

water: 50 mg/mL, clear

cation traces

Na: 13.1-18.5% (dry basis)

storage temp.

−20°C

SMILES string

[Na].OC(COP(O)(O)=O)C(O)C(O)C=O

InChI

1S/C5H11O8P.Na.H/c6-1-3(7)5(9)4(8)2-13-14(10,11)12;;/h1,3-5,7-9H,2H2,(H2,10,11,12);;

InChI key

JOSPLEVEDMZOKB-UHFFFAOYSA-N

Application

D-Arabinose 5-phosphate (D-Ara-5P) is used as a substrate to identify, differentiate and characterize enzymes such as D-arabinose 5-phosphate isomerase (KdsD) and 3-deoxy-D-manno-octulosonate 8-phosphate synthase(s) which are involved in lipopolysaccharide (LPS) biosynthesis.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louise J Gourlay et al.
Protein science : a publication of the Protein Society, 19(12), 2430-2439 (2010-10-19)
Lipopolysaccharide (LPS) biosynthesis represents an underexploited target pathway for novel antimicrobial development to combat the emergence of multidrug-resistant bacteria. A key player in LPS synthesis is the enzyme D-arabinose-5-phosphate isomerase (API), which catalyzes the reversible isomerization of D-ribulose-5-phosphate to D-arabinose-5-phosphate
Alejandra Yep et al.
Bioorganic & medicinal chemistry letters, 21(9), 2679-2682 (2011-01-18)
We explored the D-arabinose 5-phosphate isomerase (KdsD, E.C. 5.3.1.13) from Francisella tularensis, a highly infectious gram-negative pathogen that has raised concern as a potential bioweapon, as a target for the development of novel chemotherapeutics. F. tularensis KdsD was expressed in
Timothy M Allison et al.
Biochemistry, 50(18), 3686-3695 (2011-03-29)
3-Deoxy-D-manno-octulosonate 8-phosphate synthase (KDO8PS) catalyzes the reaction between three-carbon phosphoenolpyruvate (PEP) and five-carbon d-arabinose 5-phosphate (A5P), generating KDO8P, a key intermediate in the biosynthetic pathway to 3-deoxy-D-manno-octulosonate, a component of the lipopolysaccharide of the Gram-negative bacterial cell wall. Both metal-dependent
Timothy C Meredith et al.
The Journal of biological chemistry, 278(35), 32771-32777 (2003-06-14)
A gene encoding for arabinose 5-phosphate isomerase (API), which catalyzes the interconversion of d-ribulose 5-phosphate (Ru5P) and d-arabinose 5-phosphate (A5P), has been identified from the genome of Escherichia coli K-12. API is the first enzyme in the biosynthesis of 3-deoxy-d-manno-octulosonate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service