Skip to Content
Merck
All Photos(4)

Key Documents

A91906

Sigma-Aldrich

L-(+)-Arabinose

98%

Synonym(s):

(+)-Arabinose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O5
CAS Number:
Molecular Weight:
150.13
Beilstein:
1723085
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

powder

optical activity

[α]20/D +103°, c = 1 in H2O

mp

160-163 °C (lit.)

SMILES string

OC[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C5H10O5/c6-1-3(8)5(10)4(9)2-7/h1,3-5,7-10H,2H2/t3-,4-,5+/m0/s1

InChI key

PYMYPHUHKUWMLA-VAYJURFESA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-(+)-Arabinose is used as a key starting material in the total synthesis of (+)-ambruticin, zaragozic acid A, (−)-radicamine B and (+)-herbarumin I.[1][2][3][4]

Biochem/physiol Actions

L-Arabinose is the naturally occurring isomer and is a constituent of plant polysaccharides. Most bacteria contain an inducible arabinose operon that codes for a series of enzymes and transporters that allows L-arabinose to be used as the sole carbon source in microbial culture.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of (-)-radicamine B.
Gurjar, Mukund K et al.
Tetrahedron Letters, 47(39), 6979-6981 (2006)
Total synthesis of (+)-herbarumin I via intermolecular Nozaki-Hiyama-Kishi reaction.
Sabino, Adao Aparecido and Pilli, Ronaldo A
Tetrahedron Letters, 43(15), 2819-2821 (2002)
Stereoselective total synthesis of zaragozic acid A based on an acetal [1, 2] Wittig rearrangement.
Tomooka, Katsuhiko et al.
Angewandte Chemie (International Edition in English), 39(24), 4502-4505 (2000)
Total synthesis of ambruticin.
Eun Lee et al.
Angewandte Chemie (International ed. in English), 41(1), 176-178 (2002-12-20)
Catarina M S S Neves et al.
Frontiers in chemistry, 7, 459-459 (2019-07-19)
The food industry produces significant amounts of waste, many of them rich in valuable compounds that could be recovered and reused in the framework of circular economy. The development of sustainable and cost-effective technologies to recover these value added compounds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service