Skip to Content
Merck
All Photos(1)

Key Documents

A2559

Sigma-Aldrich

N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide

≥97% (HPLC), powder

Synonym(s):

Ac-DEVD-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H34N6O13
CAS Number:
Molecular Weight:
638.58
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.32

Assay

≥97% (HPLC)

form

powder

color

white

storage temp.

−20°C

SMILES string

CC(C)[C@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CC(O)=O)NC(C)=O)C(=O)N[C@@H](CC(O)=O)C(=O)Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C26H34N6O13/c1-12(2)22(26(43)30-18(11-21(38)39)24(41)28-14-4-6-15(7-5-14)32(44)45)31-23(40)16(8-9-19(34)35)29-25(42)17(10-20(36)37)27-13(3)33/h4-7,12,16-18,22H,8-11H2,1-3H3,(H,27,33)(H,28,41)(H,29,42)(H,30,43)(H,31,40)(H,34,35)(H,36,37)(H,38,39)/t16-,17-,18-,22-/m0/s1

InChI key

GGXRLUDNGFFUKI-ORGXJRBJSA-N

Gene Information

human ... CASP3(836)

Amino Acid Sequence

NAc-Asp-Glu-Val-Asp-pNA

Application

N-Acetyl-Asp-Glu-Val-Asp p-nitroanilide has been used as a chromogenic substrates for determining caspase-3 and caspase-8 activities in liver tissue homogenates and caspase 3 activity in aortic vascular smooth muscle cells and neuroblastoma cell lines.

Biochem/physiol Actions

Colorimetric substrate for caspase 3, a protease that is rapidly activated when cells are exposed to apoptotic conditions and that cleaves poly(ADP-ribose) polymerase.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nrf2 protects against 2, 3, 7, 8-tetrachlorodibenzo-p-dioxin (TCDD)-induced oxidative injury and steatohepatitis.
Lu H, et al.
Toxicology and Applied Pharmacology, 256(2), 122-135 (2011)
Juan Yao et al.
Free radical biology & medicine, 152, 659-667 (2020-01-14)
The alteration of redox homeostasis is a hallmark of cancer cells. As a critical player in regulating cellular redox signaling, thioredoxin reductase (TrxR) enzymes are increasingly recognized as attractive targets for anticancer drug development. We reported herein the natural product
Human cytomegalovirus immediate early protein 2 enhances myocardin-mediated survival of rat aortic smooth muscle cells.
Liao XH, et al.
Virus Research, 192(2), 85-91 (2014)
Familial CJD associated PrP mutants within transmembrane region induced Ctm-PrP retention in ER and triggered apoptosis by ER stress in SH-SY5Y cells.
Wang X, et al.
PLoS ONE, 6(1), e14602-e14602 (2011)
Xiangyu Wang et al.
Frontiers in cellular and infection microbiology, 7, 410-410 (2017-10-07)
Enterohemorrhagic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service