Skip to Content
Merck
All Photos(1)

Documents

185310

Sigma-Aldrich

4-Nitroaniline

≥99%

Synonym(s):

1-Amino-4-nitrobenzene, 4-Amino-1-nitrobenzene, 4-Nitro-1-aminobenzene, 4-Nitrobenzenamine, p-Aminonitrobenze

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NH2
CAS Number:
Molecular Weight:
138.12
Beilstein:
508690
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39032009
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

powder or crystals

bp

260 °C/100 mmHg (lit.)

mp

146-149 °C (lit.)

solubility

ethanol: soluble 25 mg/mL, slightly hazy, yellow to brownish-yellow
water: slightly soluble

SMILES string

Nc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H6N2O2/c7-5-1-3-6(4-2-5)8(9)10/h1-4H,7H2

InChI key

TYMLOMAKGOJONV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Nitroaniline forms molecular adducts with 4-amino benzoic acid. It reacts with nitrite ion in hydrochloric acid medium to form 4-nitrophenyldiazonium chloride, which couples with naphth-1-ol in alkaline medium to give a purple azo dye. Photocatalytic degradation of 4-nitroaniline in the presence of TiO2 suspensions in a batch and continuous annular reactor has been studied.

Application

4-Nitroaniline can be used to prepare:
  • p

  • -Phenylenediamine using various catalytic systems.
  • Poly(4-nitroaniline) thin films for optoelectronic applications.
  • An adduct with picric acid.
  • A crystalline N-(4-nitrophenyl)-β-alanine by reacting with acrylic acid.

Product of chromogenic reactions.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

415.4 °F - closed cup

Flash Point(C)

213.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal and molecular structure of N-(4-nitrophenyl)-?-alanine-Its vibrational spectra and theoretical calculations
Marchewka MK, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 79(4), 758-766 (2011)
Extraction-spectrophotometric determination of sub-microgram amounts of nitrite using 4-nitroaniline and naphth-1-ol.
Baveja AK, et al.
Analyst, 106(1266), 955-959 (1981)
A simple and highly efficient route for the preparation of p-phenylenediamine by reducing 4-nitroaniline over commercial CdS visible light-driven photocatalyst in water
Wu W, et al.
Green Chemistry, 14(6), 1705-1709 (2012)
4-Nitroaniline-picric acid (2/1)
Li Y-J
Acta Crystallographica Section E, Structure Reports Online, 65(10), o2566-o2566 (2009)
Facile synthesis of spin-coated poly (4-nitroaniline) thin film: Structural and optical properties
Abed-Elmageed AAI, et al.
Optical Materials, 109(6), 110378-110378 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service