Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
UNSPSC Code:
12161503
EC Number:
247-454-8
NACRES:
NA.84
Pricing and availability is not currently available.
Quality Segment
usage
sufficient for 1,000 multiwell tests, sufficient for 100 standard tests (2 ml)
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... CASP3(836)
General description
The Caspase 3 Fluorometric Assay Kit is based on the hydrolysis of acetyl Asp-Glu-Val-Asp 7-amido-4-methylcoumarin (Ac-DEVD-AMC) by caspase 3, resulting in the release of the fluorescent 7-amino-4-methylcoumarin (AMC). The excitation and emission wavelengths of AMC are 360 nm and 460 nm respectively. The concentration of the AMC released can be calculated from a calibration curve prepared with AMC standards (AMC standard included with the kit).
Application
The Caspase 3 Fluorimetric Assay Kit is designed for the detection and measurement of caspase 3 activity in cell extracts or purified preparations of caspase 3.
Other Notes
One unit of caspase 3 will cleave 1.0 μmol of Ac-DEVD per min at pH 7.4 at 25 °C.
1 of 1
This Item | |||
|---|---|---|---|
| Gene Information human ... CASP3(836) | Gene Information human ... CASP3(836) | Gene Information human ... CASP3(836) | Gene Information human ... CASP3(836) |
| Quality Level 200 | Quality Level 300 | Quality Level 100 | Quality Level 100 |
| usage sufficient for 1,000 multiwell tests, sufficient for 100 standard tests (2 ml) | usage sufficient for 1,000 multiwell tests (in 96-well multiwell plates), sufficient for 100 standard tests (1 ml) | usage - | usage - |
| storage temp. −20°C | storage temp. −20°C | storage temp. - | storage temp. - |
| shipped in dry ice | shipped in dry ice | shipped in dry ice | shipped in dry ice |
Kit Components Only
Product No.
Description
- Water (17-18 megaOhm) 125 mL
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk
WGK 3
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Global Trade Item Number
| SKU | GTIN |
|---|---|
| CASP3F-1KT | 04061833531594 |




