Skip to Content
Merck
All Photos(3)

Key Documents

A2251

Sigma-Aldrich

Acetyl-β-methylcholine chloride

≥98% (TLC), powder

Synonym(s):

(2-Acetoxypropyl)trimethylammonium chloride, Methacholine chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH(CH3)CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
195.69
Beilstein:
4162373
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

mp

171-173 °C (lit.)

solubility

H2O: 50 mg/mL (solutions undergo rapid, pH-dependent decomposition at pH > 6.)

storage temp.

−20°C

SMILES string

[Cl-].CC(C[N+](C)(C)C)OC(C)=O

InChI

1S/C8H18NO2.ClH/c1-7(11-8(2)10)6-9(3,4)5;/h7H,6H2,1-5H3;1H/q+1;/p-1

InChI key

JHPHVAVFUYTVCL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Acetyl-β-methylcholine (methacholine), an analog of acetylcholine, is use as a receptor agonist to study processes such as airway hyperresponsiveness associated with asthma.

Biochem/physiol Actions

Metabolically stable analog of acetylcholine; muscarinic agonist. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J G Martin et al.
Canadian journal of physiology and pharmacology, 70(4), 597-601 (1992-04-01)
Airway responsiveness to methacholine and other bronchoconstrictors is highly variable within and among species. The aim of the experiments in this report was to evaluate the importance of the quantity of airway smooth muscle as a determinant of intra- and
Bing Han et al.
PloS one, 6(2), e17236-e17236 (2011-03-02)
The great advances of nanomaterials have brought out broad important applications, but their possible nanotoxicity and risks have not been fully understood. It is confirmed that exposure of environmental particulate matter (PM), especially ultrafine PM, are responsible for many lung
Mei Hu et al.
International journal of clinical and experimental pathology, 8(11), 14171-14179 (2016-01-30)
The typical pathological features of asthma are airway remodeling and airway hyperresponsiveness (AHR). KyoT2, a negative modulator of Notch signaling, has been linked to asthma in several previous studies. However, whether KyoT2 is involved in the regulation of airway remodeling
Mina Youssef et al.
The Journal of pharmacology and experimental therapeutics, 373(3), 476-487 (2020-04-11)
Zona pellucida binding protein 2 (Zpbp2) and ORMDL sphingolipid biosynthesis regulator 3 (Ormdl3), mapped downstream of Zpbp2, were identified as two genes associated with airway hyper-responsiveness (AHR). Ormdl3 gene product has been shown to regulate the biosynthesis of ceramides. Allergic
Rifat A Chaudry et al.
Thorax, 69(6), 580-585 (2014-02-14)
Asthma and airway hyper-responsiveness are reportedly more common in children with sickle cell disease (SCD). To determine airway responsiveness, airway inflammation and clinical features of asthma in SCD. A prospective, single-centre study of 50 SCD children without overt pulmonary vascular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service