Skip to Content
Merck
All Photos(5)

Key Documents

A2218

Sigma-Aldrich

L-Ascorbic acid

meets USP testing specifications

Synonym(s):

L-Threoascorbic acid, Antiscorbutic factor, Vitamin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O6
CAS Number:
Molecular Weight:
176.12
Beilstein:
84272
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.21

biological source

synthetic

Quality Level

Agency

USP/NF
meets USP testing specifications

Assay

99.0-100.5%

form

solid

technique(s)

cell culture | mammalian: suitable

pH

1.0-2.5 (25 °C, 176 g/L in water)

mp

190-194 °C (dec.)

solubility

water: soluble 176 g/L at 20 °C

application(s)

pharmaceutical (small molecule)

SMILES string

OC([C@]([C@@H](O)CO)([H])O1)=C(O)C1=O

InChI

1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-10H,1H2/t2-,5+/m0/s1

InChI key

CIWBSHSKHKDKBQ-JLAZNSOCSA-N

Gene Information

human ... SLC23A2(9962)

Looking for similar products? Visit Product Comparison Guide

General description

L-ascorbic acid (AA) is a naturally occurring C vitamer. It is soluble in water. It is present in fruits and vegetables.

Application

L-ascorbic acid has been used as a supplement in DMEM/F12/ osteoblast induction medium. It has also been used as a component in liquid Ham′s isolation media.

Biochem/physiol Actions

L-ascorbic acid (AA) can block and treat scurvy. It possesses antioxidant properties. In the fermentation of wine, AA acid serve as a food additive and antioxidant.
Ascorbic Acid, also known as Vitamin C, is a six-carbon lactone produced by plants and some animal species but not by humans and other primates. Ascorbic acid functions as an enzymatic cofactor for multiple enzymes, serving as an electron donor for monooxygenases and dioxygenases. Ascorbic acid also functions as a powerful antioxidant, particularly in regards to reactive oxygen species.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1, 3, 2-Dioxathiolane oxides: epoxide equivalents and versatile synthons
Lohray BB and Bhushan V
Advances in Heterocyclic Chemistry, 68, 90-181 (1997)
Analysis of vitamins by liquid chromatography
Liquid Chromatography, 571-615 (2017)
Biological characteristics of muscle-derived satellite cells isolated from rats at different postnatal days
Yu R, et al.
Cytotechnology, 67(3), 397-408 (2015)
Compositional analysis of foods
Liquid Chromatography (2013)
An improved protocol for the isolation and cultivation of embryonic mouse myocytes
Rodgers LS, et al.
Cytotechnology, 59(2), 93-102 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service