Skip to Content
Merck
All Photos(1)

Documents

E0377

Sigma-Aldrich

D-Erythrose 4-phosphate sodium salt

≥50% (TLC)

Synonym(s):

4-Phospho-D-erythrose sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8NaO7P
CAS Number:
Molecular Weight:
222.07
UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

Assay

≥50% (TLC)

form

powder

color

white to off-white

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

−20°C

SMILES string

[Na+].[H]C(=O)[C@H](O)[C@H](O)COP(O)([O-])=O

InChI

1S/C4H9O7P.Na/c5-1-3(6)4(7)2-11-12(8,9)10;/h1,3-4,6-7H,2H2,(H2,8,9,10);/q;+1/p-1/t3-,4+;/m0./s1

InChI key

KKDBADMPNGAKHM-RFKZQXLXSA-M

Looking for similar products? Visit Product Comparison Guide

Application

D-Erythrose 4-phosphate (E4P) is used as a substrate to identify, differentiate and characterize 2-keto-3-deoxy-6-phosphogalactonate (KDPGal) aldolase(s) and 3-deoxy-D-arabino-heptulosonate 7-phosphate (DAH7P) synthase(s) that initiate the shikimate pathway in microorganisms and plants. It may be used as a substrate to identify, differentiate and characterize glyceraldehyde-3-phosphate dehydrogenase(s) (GAPDH) involved in surface antigen and virulence factor development in pathogenic microorganisms. E4P may be used in malaria research as an inhibitor of the PfPdx1 component of the PLP (pyridoxal 5′-phosphate) synthase enzymatic complex responsible for the de novo synthesizes vitamin B6 in Plasmodium falciparum.

Biochem/physiol Actions

Erythrose 4-phosphate is an intermediate in the penntose phosphate pathway, as well as the biosynthesis of phenylalanine, tyrosine, tryptophan and metabolism of vitamin B6.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bouchra Elkhalfi et al.
Protein expression and purification, 89(2), 146-155 (2013-03-20)
The gammaproteobacterium Pseudomonas syringae pv. tomato DC3000 is the causal agent of bacterial speck, a common disease of tomato. The mode of infection of this pathogen is not well understood, but according to molecular biological, genomic and proteomic data it
Shaun B Reeksting et al.
The Biochemical journal, 449(1), 175-187 (2012-10-09)
Malaria tropica is a devastating infectious disease caused by Plasmodium falciparum. This parasite synthesizes vitamin B6 de novo via the PLP (pyridoxal 5'-phosphate) synthase enzymatic complex consisting of PfPdx1 and PfPdx2 proteins. Biosynthesis of PLP is largely performed by PfPdx1
Celia J Webby et al.
The Biochemical journal, 390(Pt 1), 223-230 (2005-04-28)
DAH7P (3-Deoxy-D-arabino-heptulosonate 7-phosphate) synthase catalyses the condensation reaction between phosphoenolpyruvate (PEP) and D-erythrose 4-phosphate (E4P) as the first committed step in the biosynthesis of aromatic compounds in plants and micro-organisms. Previous work has identified two families of DAH7P synthases based
Scott R Walker et al.
Bioorganic & medicinal chemistry letters, 21(17), 5092-5097 (2011-04-16)
3-Deoxy-d-arabino-heptulosonate 7-phosphate (DAH7P) synthase catalyses the first step of the shikimate pathway for the biosynthesis of aromatic compounds. Enzymes of this pathway have been identified as potential targets for drug design. The reaction catalysed by DAH7P synthase is an aldol
J Bongaerts et al.
Metabolic engineering, 3(4), 289-300 (2001-10-26)
Metabolic engineering to design and construct microorganisms suitable for the production of aromatic amino acids and derivatives thereof requires control of a complicated network of metabolic reactions that partly act in parallel and frequently are in rapid equilibrium. Engineering the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service