Skip to Content
Merck
All Photos(1)

Key Documents

444100

Millipore

D-Mannose-6-phosphate, Disodium Salt

Synonym(s):

D-Mannose-6-phosphate, Disodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11O9P · 2Na
CAS Number:
Molecular Weight:
304.10
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79
Pricing and availability is not currently available.

Assay

≥97% dry basis (enzymatic)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<0.1% fructose-1,6-diphosphate, fructose-6-phosphate, glucose-6-phosphate

color

white

solubility

water: soluble

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[Na+].[P](=O)([O-])([O-])OCC1OC(C(C(C1O)O)O)O

InChI

1S/C6H13O9P.2Na/c7-3-2(1-14-16(11,12)13)15-6(10)5(9)4(3)8;;/h2-10H,1H2,(H2,11,12,13);;/q;2*+1/p-2

InChI key

CHIBNKGHYAQTQY-UHFFFAOYSA-L

Related Categories

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service