Skip to Content
Merck
All Photos(1)

Key Documents

A5295

Sigma-Aldrich

Anti-Bovine IgG (whole molecule)−Peroxidase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Rabbit Anti-Bovine IgG (whole molecule)−HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

bovine

technique(s)

direct ELISA: 1:20,000
dot blot: 1:80,000 (chemiluminescent)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

IgG (immunoglobulin G) contributes to 10-20% of plasma protein and is regarded as one of the most predominant serum protein. It consists of four subclasses: IgG1, IgG2, IgG3 and IgG4. The IgG structure possesses four polypeptide chains containing two identical γ heavy (H) chains and two identical κ or λ light (L) chains of 50 kDa and 25 kDa, respectively.

Immunogen

purified bovine IgG

Application

Anti-Bovine IgG (whole molecule)-Peroxidase antibody produced in rabbit has been used in:
  • enzyme-linked immunosorbent assay (ELISA)
  • bovine viral diarrhea virus (BVDV) plaque
  • focus-forming assays

Biochem/physiol Actions

Bovine IgGs are glycoprotein antibodies that regulate immune responses in herds. These antibodies inhibit TLR5 activation upon immunization with native H7 flagellin. Bovine IgG levels can be used for the detection of Johne′s disease and milk adulteration.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 0.05% MIT.

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Garza-Cuartero et al.
Vaccine, 36(7), 958-968 (2018-01-27)
Fasciola hepatica infection causes important economic losses in livestock and food industries around the world. In the Republic of Ireland F. hepatica infection has an 76% prevalence in cattle. Due to the increase of anti-helminthic resistance, a vaccine-based approach to
Rafaella P M Guimarães-Peixoto et al.
PLoS neglected tropical diseases, 12(4), e0006371-e0006371 (2018-04-13)
Bovine cysticercosis is a worldwide distributed zoonosis caused by the larval form of Taenia saginata present in bovine muscles. The diagnosis is based on the postmortem inspection at slaughterhouses and consists of the macroscopic visualization of lesions caused by cysticercosis
Infectious bovine viral diarrhea virus (strain NADL) RNA from stable cDNA clones: a cellular insert determines NS3 production and viral cytopathogenicity
Mendez E, et al.
Journal of virology, 72(6), 4737-4745 (1998)
Karukriti Kaushik Ghosh et al.
Applied and environmental microbiology, 84(6) (2017-12-23)
In this study, the effect of the host stress hormone catecholamine on Leptospira gene transcripts encoding outer membrane proteins was investigated. There was no impact of catecholamine supplementation on the in vitro growth pattern of Leptospira interrogans; however, 7 genes
Davita Pillay et al.
PloS one, 8(10), e78565-e78565 (2013-11-10)
Diagnosis of African animal trypanosomosis is vital to controlling this severe disease which hampers development across 10 million km(2) of Africa endemic to tsetse flies. Diagnosis at the point of treatment is currently dependent on parasite detection which is unreliable

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service