Skip to Content
Merck
All Photos(1)

Documents

12-348

Sigma-Aldrich

Goat Anti-Rabbit IgG Antibody, HRP-conjugate

1 mg/mL, Upstate®

Synonym(s):

Anti-rabbit IgG

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Upstate®

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

Related Categories

General description

Immunoglobulin G (IgG), is one of the most abundant proteins in human serum with normal levels between 8-17 mg/mL in adult blood. IgG is important for our defence against microorganisms and the molecules are produced by B lymphocytes as a part of our adaptive immune response. The IgG molecule has two separate functions; to bind to the pathogen that elicited the response and to recruit other cells and molecules to destroy the antigen. The variability of the IgG pool is generated by somatic recombination and the number of specificities in an individual at a given time point is estimated to be 1011 variants.

Specificity

Specific for rabbit IgG heavy and light chains.

Immunogen

Highly purified whole rabbit IgG.

Application

Goat Anti-Rabbit IgG Antibody, HRP-conjugate detects level of Rabbit IgG & has been published & validated for use in WB, IH, ELISA.

Quality

Evaluated by western blot.

Western Blot Analysis: Suitable for western blotting (dot blot), ELISA and immunohistochemistry. A working dilution of 1:15,000 to 1:60,000 is suggested for ELISA. A working dilution of 1:2,000 to 1:6,000 is suggested for western blotting procedures. A working dilution of 1:500 to 1:3,000 is suggested for immunohistochemistry. Optimal antibody dilution for other applications should be determined by the researcher.

Physical form

Goat polyclonal conjugated with horseradish peroxidase IgG in buffer containing 50% storage buffer 0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 10 mg/mL BSA, 0.01% (w/v) gentamicin sulfate and 50% glycerol.
Rehydration: Add 500 µL sterile, distilled water containing 50% glycerol, to make a 1 mg/mL stock solution.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation-induced cell death and total Akt content of granulocytes show a biphasic course after low-dose radiation.
Gaipl, U S, et al.
Autoimmunity, 42, 340-342 (2009)
Geovanny I Nic-Can et al.
Methods in molecular biology (Clifton, N.J.), 877, 313-324 (2012-05-23)
Epigenetics includes DNA methylation and histones posttranslational modifications such as methylation, acetylation, phosphorylation among others. One of the most abundant modifications in histone tail is the methylation. It has been found that the methylation pattern in the histone H3 may
Kuo-Sheng Hsu et al.
The Journal of biological chemistry, 288(35), 25375-25386 (2013-07-19)
Cytokine modulation of the endothelium is considered an important contributor to the inflammation response. TNFα is an early response gene during the initiation of inflammation. However, the detailed mechanism by which TNFα induces proinflammatory gene expression is not completely understood.
Caroline M Forrest et al.
Brain research, 1504, 1-15 (2013-01-29)
Glutamate receptors sensitive to N-methyl-d-aspartate (NMDA) are important in early brain development, influencing cell proliferation and migration, neuritogenesis, axon guidance and synapse formation. The kynurenine pathway of tryptophan metabolism includes an agonist (quinolinic acid) and an antagonist (kynurenic acid) at
Maj Sundbom et al.
BMC pharmacology, 8, 3-3 (2008-02-14)
A substantial body of evidence indicates that reduced plasma adiponectin levels may be key in the development of insulin resistance, type 2 diabetes and the metabolic syndrome. Glucocorticoids decrease the levels of adiponectin in animals and humans. Cortisone is transformed

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service