Skip to Content
Merck
All Photos(2)

Documents

A8786

Sigma-Aldrich

Anti-Mouse IgM (μ-chain specific)–Peroxidase antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Goat Anti-Mouse IgM (μ-chain specific)–HRP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

direct ELISA: 1:10,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

IgM is a glycoprotein antibody that regulates humoral immune responses. Mouse IgM is involved in modulating B cell memory . This antibody isotype has also been implicated in the development of autoimmune responses associated with the pathogenesis of type 1 diabetes in mice . Anti-Mouse IgM (μ-chain specific)-Peroxidase antibody is specific for mouse IgM when tested against purified mouse IgA, IgM, and IgG.

Immunogen

Purified mouse IgM

Application

Anti-Mouse IgM (μ-chain specific)-Peroxidase antibody produced in goat has also been used in:
  • western blotting
  • enzyme linked immunosorbent assay (ELISA)
  • immunohistochemistry

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4 containing 1% bovine serum albumin with preservative.

Preparation Note

Prepared using the periodate method described by Wilson, M.B., and Nakane, P.K., in Immunofluorescence and Related Staining Techniques, Elsevier/North Holland Biomedical Press, Amsterdam, p215 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Krzysztof Wandzik et al.
Development, growth & differentiation, 51(6), 555-565 (2009-08-01)
We investigated the remodeling of iron metabolism during megakaryocytic development of K562 cells. Differentiation was successfully verified by increase of the megakaryocytic marker CD61 and concomitant decrease of the erythroid marker γ-globin. The reduction of erythroid properties was accompanied by
Yan Wang et al.
The Journal of biological chemistry, 291(19), 10046-10057 (2016-03-10)
The partially de-N-acetylated poly-β-1,6-N-acetyl-d-glucosamine (dPNAG) polymer serves as an intercellular biofilm adhesin that plays an essential role for the development and maintenance of integrity of biofilms of diverse bacterial species. Translocation of dPNAG across the bacterial outer membrane is mediated
Michael Wetter et al.
PloS one, 7(9), e45609-e45609 (2012-10-03)
The Vi capsular polysaccharide (CPS) of Salmonella enterica serovar Typhi, the cause of human typhoid, is important for infectivity and virulence. The Vi biosynthetic machinery is encoded within the viaB locus composed of 10 genes involved in regulation of expression
Enhanced humoral immunity in mice lacking CB1 and CB2 receptors (Cnr1-/-/Cnr2-/- mice) is not due to increased splenic noradrenergic neuronal activity
Simkins T, et al.
Journal of Neuroimmune Pharmacology, 9(4), 544-557 (2014)
Asfia Qureshi et al.
Mycopathologia, 173(5-6), 419-425 (2011-10-06)
We have developed an ELISA to determine the value of anti-glucosylceramide antibody for the prediction of disseminated cryptococcosis in immunocompromised subjects and performed a clinical prospective study at the Medical University of South Carolina. The study enrolled a total of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service