Skip to Content
Merck
All Photos(1)

Key Documents

A0138

Sigma-Aldrich

Aflatoxin G1

from Aspergillus flavus

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12O7
CAS Number:
Molecular Weight:
328.27
Beilstein:
1299768
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

Aspergillus flavus

Quality Level

form

powder

storage temp.

2-8°C

SMILES string

COc1cc2OC3OC=CC3c2c4OC(=O)C5=C(CCOC5=O)c14

InChI

1S/C17H12O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h3,5-6,8,17H,2,4H2,1H3

InChI key

XWIYFDMXXLINPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxins are hepatotoxic metabolites, that are produced by molds, such as, Aspergillus flavus and Aspergillus parasiticus. They are naturally occurring poisonous carcinogens and the most widely known mycotoxins. They are associated with liver cancer and immune system suppression.

Application

Aflatoxin G1 from Aspergillus flavus has been used as a standard for the determination of aflatoxin in various samples.

Biochem/physiol Actions

Hepatocarcinogen. Food contaminant produced by Aspergillus flavus, a common soil fungus.

Warning

Possibly carcinogenic.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Carc. 1B - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Review of past and present research on Aflatoxin in Uganda
Sittig's Handbook of Toxic and Hazardous Chemicals and Carcinogens (2008)
Validation of simultaneous analysis method for determination of aflatoxins in olive oil by high performance liquid chromatographyfluorescence detector
Nabizadeh S, et al.
Journal of Food Safety and Hygiene, 1(2), 63-68 (2015)
The use of regenerated immunoaffinity columns for aflatoxins B1, B2, G1 and G2 in peanut confection
Iha Mh, et al.
Journal of Chromatography A, 1483, 1-7 (2017)
Review of past and present research on Aflatoxin in Uganda
Kaaya NA and Warren HL
African Journal of Food, Agriculture, Nutrition and Development, 5(1) (2005)
Sharaf S Omar et al.
Foods (Basel, Switzerland), 9(5) (2020-05-24)
In Jordan as in other worldwide countries, mycotoxins are considered a serious national problem in food supplies. As a result, almost all nations are setting and adopting different regulations targeting the control of mycotoxins levels in the domestic food supply

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service