Skip to Content
Merck
All Photos(1)

Key Documents

A0263

Sigma-Aldrich

Aflatoxin G2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Beilstein:
1692017
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

Quality Level

storage temp.

2-8°C

SMILES string

[H][C@@]12CCO[C@]1([H])Oc3cc(OC)c4C5=C(C(=O)OCC5)C(=O)Oc4c23

InChI

1S/C17H14O7/c1-20-9-6-10-12(8-3-5-22-17(8)23-10)14-11(9)7-2-4-21-15(18)13(7)16(19)24-14/h6,8,17H,2-5H2,1H3/t8-,17+/m0/s1

InChI key

WPCVRWVBBXIRMA-WNWIJWBNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aflatoxin G2 is a fungal secondary toxic metabolite synthesized by Aspergillus parasiticus, Aspergillus flavus, and Aspergillus nomius, species that are found in tropical and subtropical regions of the world. Aflatoxin Gs (AFGs) are derivatives of difuranocoumarin and contain a bifuran group linked to a coumarin nucleus and a lactone ring.

Application

Aflatoxin G2 has been used as a standard to estimate the levels of aflatoxins by immunoaffinity column cleanup in:
  • peanut confection
  • Ginseng and other roots
  • olive oil, peanut oil, and sesame oil

Biochem/physiol Actions

Aflatoxins are hepatocarcinogenic and act as food contaminants in peanuts, fruits, cereals, corn, and dry fruits.

Warning

Possibly carcinogenic.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Acute Tox. 2 Inhalation - Carc. 1B - Muta. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dipendra K Mahato et al.
Frontiers in microbiology, 10, 2266-2266 (2019-10-23)
Aflatoxins produced by the Aspergillus species are highly toxic, carcinogenic, and cause severe contamination to food sources, leading to serious health consequences. Contaminations by aflatoxins have been reported in food and feed, such as groundnuts, millet, sesame seeds, maize, wheat
Maria Helena Iha et al.
Journal of chromatography. A, 1483, 1-7 (2017-01-09)
The aim of this study was to investigate the feasibility of using multitime-regenerated immunoaffinity column (IAC) for aflatoxins B1, B2, G1 and G2 in peanut confection. After each use, the IAC was washed immediately with phosphate-buffered saline and stored for >12h
Mary Trucksess et al.
Journal of AOAC International, 89(3), 624-630 (2006-06-24)
Mycotoxins are toxic secondary metabolites produced by certain molds and are common contaminants of many important food crops, such as grains, nuts, and spices. Some mycotoxins are found in fruits, vegetables, and botanical roots. These contaminants have a broad range
Meritxell Ventura et al.
Journal of chromatography. A, 1048(1), 25-29 (2004-09-30)
An easy method for the determination of aflatoxins B1, G1, B2 and G2 in Rhammus purshiana by LC coupled to mass spectrometry has been developed. Aflatoxins were extracted with a mixture of methanol and water and then it was purified
Evaluation and improvement of extraction methods for the analysis of aflatoxins B1, B2, G1 and G2 from naturally contaminated maize
Bertuzzi T, et al.
Food Analytical Methods, 512-519 (2012)

Articles

Cancer research has revealed that the classical model of carcinogenesis, a three step process consisting of initiation, promotion, and progression, is not complete.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service