Skip to Content
Merck
All Photos(3)

Documents

208094

Sigma-Aldrich

Magnesium sulfate

anhydrous, reagent grade, ≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgSO4
CAS Number:
Molecular Weight:
120.37
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous
reagent grade

Quality Level

vapor density

<0.01 (vs air)

vapor pressure

<0.1 mmHg ( 20 °C)

Assay

≥97%

form

granular

pH

~7.9 (25 °C, 50 g/L)

SMILES string

[Mg++].[O-]S([O-])(=O)=O

InChI

1S/Mg.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2

InChI key

CSNNHWWHGAXBCP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium sulfate (MgSO4) has been used as a medium supplement in the M63 minimal medium for bacteria.
Anhydrous MgSO4 plays the role of a drying agent in the synthesis of 3,6-bis(3,4′-dihexyl-2,2′-bithiophen-5-yl)-1,2,4,5-tetrazine and organic molecules having an A–D–A (D = donor, A = acceptor) structure.
It may be used as an additional reagent in the synthesis of t-butyl esters and ethers by reacting carboxylic acid or alcohol and t-butanol in the presence of catalytic sulfuric acid.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adherent-invasive Escherichia coli Biofilm Formation Assays.
Chassaing B and Darfeuille-Michaud A.
Bio-protocol, 3(23) (2013)
Dithienosilole-bridged small molecules with different alkyl group substituents for organic solar cells exhibiting high open-circuit voltage
Ye, Dandan, et al.
Journal of Material Chemistry A, 1.26, 7622-7629 (2013)
Convenient preparations of t-butyl esters and ethers from t-butanol.
Wright SW, et al.
Tetrahedron Letters, 38(42), 7345-7348 (1997)
Donor?acceptor copolymers incorporating polybenzo [1, 2-b: 4, 5-b'] dithiophene and tetrazine for high open circuit voltage polymer solar cells
Cheng W, et al.
Organic Electronics, 14.9, 2124-2131 (2013)
Lex W Doyle
Current opinion in pediatrics, 24(2), 154-159 (2012-01-10)
Antenatal magnesium sulfate may reduce the excessive rates of cerebral palsy in survivors of very preterm birth. There are five randomized controlled trials of magnesium sulfate therapy given to the mother prior to very preterm birth which have reported neurological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service