Skip to Content
Merck
All Photos(1)

Key Documents

81028

Sigma-Aldrich

Potassium sodium tartrate solution

BioUltra, 1.5 M in H2O

Synonym(s):

L-(+)-Tartaric acid potassium Sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4KNaO6
CAS Number:
Molecular Weight:
210.16
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.26
Pricing and availability is not currently available.

product line

BioUltra

Quality Level

form

liquid

optical activity

[α]20/D +22±1°, c = 1% in H2O

concentration

1.5 M in H2O

impurities

insoluble matter, passes filter test
≤0.002% total nitrogen (N)

pH

7.0-8.5 (25 °C, 1.5 M in H2O)

anion traces

chloride (Cl-): ≤5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

1.5 M in H2O

UV absorption

λ: 260 nm Amax: 0.03
λ: 280 nm Amax: 0.025

SMILES string

[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.K.Na/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;/q;2*+1/p-2/t1-,2-;;/m1../s1

InChI key

LJCNRYVRMXRIQR-OLXYHTOASA-L

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Ledwoch et al.
Letters in applied microbiology, 68(4), 329-336 (2019-02-26)
Dry surface biofilms (DSB) harbouring pathogens are widespread in healthcare settings, are difficult to detect and are resistant to cleaning and disinfection interventions. Here, we describe a practical test protocol to palliate the lack of standard efficacy test methods for
Ryan T Terry-Lorenzo et al.
Journal of biomolecular screening, 20(10), 1218-1231 (2015-08-25)
Genome-wide association studies have linked polymorphisms in the gene G72 to schizophrenia risk in several human populations. Although controversial, biochemical experiments have suggested that the mechanistic link of G72 to schizophrenia is due to the G72 protein product, pLG72, exerting

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service