Skip to Content
Merck
All Photos(1)

Documents

32312

Sigma-Aldrich

Potassium sodium tartrate tetrahydrate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (perchloric acid titration)

Synonym(s):

L(+)-Tartaric acid potassium Sodium salt, Rochelle salt, Seignette salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KOCOCH(OH)CH(OH)COONa · 4H2O
CAS Number:
Molecular Weight:
282.22
Beilstein:
6113568
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Quality Level

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

Assay

≥99.5% (perchloric acid titration)

impurities

≤0.001% total nitrogen (N)

pH

7.0-8.5 (20 °C, 5%)

anion traces

chloride (Cl-): ≤5 mg/kg
phosphate (PO43-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤40 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Pb: ≤2 mg/kg
Zn: ≤5 mg/kg

SMILES string

O.O.O.O.[Na+].[K+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.K.Na.4H2O/c5-1(3(7)8)2(6)4(9)10;;;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;4*1H2/q;2*+1;;;;/p-2/t1-,2-;;;;;;/m1....../s1

InChI key

VZOPRCCTKLAGPN-ZFJVMAEJSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium sodium tartrate tetrahydrate is a double salt of tartaric acid that can be used to prevent the precipitation in alkaline medium. It can also be used as a reagent in the determination of nitrilase activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of Commercial Nitrilase Hydrolysis on Acrylic Fabrics
Kim H, et al.
Textile Research Journal, 18, 889-896 (2016)
Potassium sodium (2R, 3R)-tartrate tetrahydrate: the paraelectric phase of Rochelle salt at 105K.
Gorbitz CH and Sagstuen E.
Acta Crystallographica Section E, Structure Reports Online, 64(4), 507-508 (2008)
Nandini P Shetty et al.
Journal of experimental botany, 60(15), 4287-4300 (2009-11-03)
The accumulation of the pathogenesis-related (PR) proteins beta-1,3-glucanase and chitinase and structural defence responses were studied in leaves of wheat either resistant or susceptible to the hemibiotrophic pathogen Septoria tritici. Resistance was associated with an early accumulation of beta-1,3-glucanase and
Ghina Ali et al.
Biotechnology progress, 31(4), 883-889 (2015-04-29)
The immobilization of pullulanase from Klebsiella pneumoniae by grafting was investigated. Pullulanase was linked after activation of alginate via a covalent bond between the amine groups of the enzyme and the carboxylic acid groups of alginate. The immobilization yield was
Catarina M S S Neves et al.
Physical chemistry chemical physics : PCCP, 17(47), 32044-32052 (2015-11-18)
Due to scarce available experimental data, as well as due to the absence of predictive models, the influence of salts on the solubility of ionic liquids (ILs) in water is still poorly understood. To this end, this work addresses the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service