Skip to Content
Merck
All Photos(1)

Key Documents

78848

Sigma-Aldrich

2-Ethylhexyl 4-methoxycinnamate

98%

Synonym(s):

4-Methoxycinnamic acid 2-ethylhexyl ester, OMC, Octyl methoxycinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O3
CAS Number:
Molecular Weight:
290.40
Beilstein:
5946632
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.25

Assay

98% (GC)
98%

form

liquid

refractive index

n20/D 1.545

SMILES string

CCCCC(CC)COC(=O)\C=C\c1ccc(OC)cc1

InChI

1S/C18H26O3/c1-4-6-7-15(5-2)14-21-18(19)13-10-16-8-11-17(20-3)12-9-16/h8-13,15H,4-7,14H2,1-3H3/b13-10+

InChI key

YBGZDTIWKVFICR-JLHYYAGUSA-N

Looking for similar products? Visit Product Comparison Guide

General description

OMC provides protection against UV induced cyclobutane pyrimidine dimer (CDP) formation, filtering UVB.

Application


  • Simultaneous electrochemical determination of three sunscreens using cetyltrimethylammonium bromide.: This study presents an electrochemical method for the simultaneous determination of sunscreens, including 2-Ethylhexyl 4-methoxycinnamate, using cetyltrimethylammonium bromide, highlighting the importance of analytical techniques in biochemistry (Cardoso et al., 2008).

  • Relationship between the ability of sunscreens containing 2-ethylhexyl-4′-methoxycinnamate to protect against UVR-induced inflammation, depletion of epidermal Langerhans (Ia+) cells and suppression of alloactivating capacity of murine skin in vivo.: This study evaluates the protective effects of sunscreens with 2-Ethylhexyl 4-methoxycinnamate against UV radiation-induced skin changes, contributing to the understanding of immune responses in dermatological biochemistry (Walker et al., 1994).

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

379.4 °F

Flash Point(C)

193 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nur Duale et al.
Toxicological sciences : an official journal of the Society of Toxicology, 114(2), 272-284 (2010-01-15)
Octyl methoxycinnamate (OMC) is one of the most widely used sunscreen ingredients. To analyze biological effects of OMC, an in vitro approach was used implying ultraviolet (UV) exposure of two human cell lines, a primary skin fibroblast (GM00498) and a
Juan L Benedé et al.
Talanta, 147, 246-252 (2015-11-26)
In this work, a new approach that combines the advantages of stir bar sorptive extraction (SBSE) and dispersive solid phase extraction (DSPE), i.e. stir bar sorptive-dispersive microextraction (SBSDµE), is employed as enrichment and clean-up technique for the sensitive determination of
Wenlan Song et al.
Journal of chromatography. A, 1384, 28-36 (2015-02-11)
A novel zinc-zinc oxide (Zn-ZnO) nanosheets coating was directly fabricated on an etched stainless steel wire substrate as solid-phase microextraction (SPME) fiber via previous electrodeposition of robust Zn coating. The scanning electron micrograph of the Zn-ZnO nanosheets coated fiber exhibits
Yida Zhang et al.
Analytica chimica acta, 876, 55-62 (2015-05-23)
A novel cedar-like Au nanoparticles (AuNPs) coating was fabricated on an etched stainless steel (SS) wire by direct chemical deposition and used as an efficient and unbreakable solid phase microextraction (SPME) fiber. The etched SS wire offers a rough surface
Sean Ekins et al.
Drug metabolism and disposition: the biological fate of chemicals, 38(12), 2302-2308 (2010-09-17)
Drug-induced liver injury (DILI) is one of the most important reasons for drug development failure at both preapproval and postapproval stages. There has been increased interest in developing predictive in vivo, in vitro, and in silico models to identify compounds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service