Skip to Content
Merck
All Photos(3)

Documents

437166

Sigma-Aldrich

2-Phenyl-5-benzimidazolesulfonic acid

96%

Synonym(s):

Ensulizole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10N2O3S
CAS Number:
Molecular Weight:
274.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

mp

>300 °C (lit.)

solubility

ethanol and water: soluble (as sodium salt)

SMILES string

OS(=O)(=O)c1ccc2[nH]c(nc2c1)-c3ccccc3

InChI

1S/C13H10N2O3S/c16-19(17,18)10-6-7-11-12(8-10)15-13(14-11)9-4-2-1-3-5-9/h1-8H,(H,14,15)(H,16,17,18)

InChI key

UVCJGUGAGLDPAA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenyl-5-benzimidazolesulfonic acid has been reported as UV filter. It is a xenobiotic compound detected in in gray water from houses and effluents of the biological treatment systems (aerobic, anaerobic, and combined anaerobic+aerobic). It is a micropollutant of personal care products and its removal from aerobically treated grey water by ozonation and adsorption onto activated carbon has been reported.

Application

2-Phenyl-5-benzimidazolesulfonic acid was used for the fabrication of biomimetic sensor for determination of 4-methylbenzylidene camphor (4-MBC), an ultraviolet (UV) radiation protector.
UV absorber

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Hernández-Leal et al.
Water research, 45(9), 2887-2896 (2011-04-02)
Ozonation and adsorption onto activated carbon were tested for the removal micropollutants of personal care products from aerobically treated grey water. MilliQ water spiked with micropollutants (100-1600 μgL(-1)) was ozonated at a dosing rate of 1.22. In 45 min, this
Lucía Hernández Leal et al.
Environmental science & technology, 44(17), 6835-6842 (2010-08-05)
Eighteen selected xenobiotics related to personal care and household chemicals (UV-filters, fragrances, preservatives, biocides, surfactants) were measured in gray water from 32 houses and in effluents of three different biological treatment systems (aerobic, anaerobic, and combined anaerobic+aerobic). All selected xenobiotics
Application of a biomimetic sensor based on iron phthalocyanine chloride: 4-methylbenzylidene-camphor detection.
Boni AC, et al.
Journal of the Brazilian Chemical Society, 21(7), 1377-1383 (2010)
Ashley R Heurung et al.
Dermatitis : contact, atopic, occupational, drug, 25(6), 289-326 (2014-11-11)
Sunscreen is a key component in the preventive measures recommended by dermatologists and public health campaigns aimed at reducing sunburn, early skin aging, and skin cancer. To maximize compliance, adverse reactions to sunscreens should be minimized. Although inactive ingredients cause
Murilo L Bello et al.
PloS one, 10(3), e0121110-e0121110 (2015-03-25)
Layered drug delivery carriers are current targets of nanotechnology studies since they are able to accommodate pharmacologically active substances and are effective at modulating drug release. Sodium montmorillonite (Na-MMT) is a clay that has suitable properties for developing new pharmaceutical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service