Skip to Content
Merck
All Photos(1)

Documents

437379

Sigma-Aldrich

2-Ethylhexyl salicylate

99%

Synonym(s):

Octisalate, Octyl salicylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)C6H4CO2CH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
250.33
Beilstein:
2730664
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

liquid

refractive index

n20/D 1.502 (lit.)

bp

189-190 °C/21 mmHg (lit.)

density

1.014 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)c1ccccc1O

InChI

1S/C15H22O3/c1-3-5-8-12(4-2)11-18-15(17)13-9-6-7-10-14(13)16/h6-7,9-10,12,16H,3-5,8,11H2,1-2H3

InChI key

FMRHJJZUHUTGKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexyl salicylate(EHS) is an organic ultraviolet(UV) absorber that can be used as a photostable ingredient in cosmetic formulations. It shows an absorption spectra in the range of 280-320 nm in the UV region.

Application

EHS is a salicylic ester which can be used as an UV filter in sunscreen based creams.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

323.6 °F - closed cup

Flash Point(C)

162 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A pendant peptide endows a sunscreen with water-resistance.
Ellison AJ and Raines RT
Organic & Biomolecular Chemistry, 16(39), 7139-7142 (2018)
Optical and electron paramagnetic resonance studies of the excited triplet states of UV-B absorbers: 2-ethylhexyl salicylate and homomenthyl salicylate.
Sugiyama K, et al.
Photochemical & Photobiological Sciences : Official Journal of the European Photochemistry Association and the European Society for Photobiology, 14(9), 1651-1659 (2015)
P Clarys et al.
Journal of dermatological science, 27(2), 77-81 (2001-09-05)
Temperature influences the stratum corneum adsorption of several topically applied compounds. This study was designed to evaluate the influence of the temperature on the stratum corneum adsorption of 3 UV filters. The UV filters were solubilized in two vehicles, an
M McVean et al.
Carcinogenesis, 18(8), 1617-1622 (1997-08-01)
Ultraviolet B (UVB, 290-320 nm) exposure results in a variety of cellular insults including induction of cyclobutane pyrimidine dimers in DNA. Accumulation of these lesions can lead to mutations in critical genes and contribute to the development of nonmelanoma skin
Beatriz Albero et al.
Journal of chromatography. A, 1248, 9-17 (2012-06-12)
The determination of organic contaminants in soil is a real challenge due to the large number of these compounds with quite different physico-chemical properties. In the present work, an analytical method was developed for the simultaneous determination in soil of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service