Skip to Content
Merck
All Photos(1)

Key Documents

75259

Sigma-Aldrich

Tris(2-carboxyethyl)phosphine hydrochloride

BioUltra, ≥98% (NMR)

Synonym(s):

TCEP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15O6P · HCl
CAS Number:
Molecular Weight:
286.65
Beilstein:
3724376
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

Assay

≥98% (NMR)

form

crystals

reaction suitability

reagent type: reductant

loss

≤1.0% loss on drying

pH

1.2-1.5

solubility

H2O: 1 M, clear, colorless (no residue)

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.05 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

0.1 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.03

storage temp.

2-8°C

SMILES string

Cl[H].OC(=O)CCP(CCC(O)=O)CCC(O)=O

InChI

1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H

InChI key

PBVAJRFEEOIAGW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tris(2-carboxyethyl)-phosphine hydrochloride (TCEP) is a strong reducing agent that is stable in an aqueous solution. It readily and stoichiometrically reduces disulfides with high specificity. TCEP exhibits reducing activity at pH 1.5 to pH 9.0 and reduces dehydroascorbic acid (DHA) at pH 4.

Application

Tris(2-carboxyethyl)phosphine hydrochloride has been used:
  • as a component of IRE1 dialysis buffer, RNA cleavage buffer, and in in vitro cleavage assay of in vitro transcribed RNA
  • in reducing sulfhydryl groups to obtain total thiol fraction
  • to reduce protein samples

Water soluble reagent, used for selective reduction of disulfides. More stable than DTT and useful in mass spectrometry applications.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Elif Karagöz et al.
Bio-protocol, 9(14), e3307-e3307 (2019-07-20)
The kinase/RNase IRE1 is a key effector of the cellular response to endoplasmic reticulum stress. The RNase activity of IRE1 can be measured in cells or in the test tube. Here we describe a protocol for the in vitro cleavage
Yasunori Sato et al.
Biological & pharmaceutical bulletin, 33(3), 364-369 (2010-03-02)
Ascorbic acid (AA) has a strong anti-oxidant function evident as its ability to scavenge superoxide radicals in vitro. Moreover, AA is an essential ingredient for post-translational proline hydroxylation of collagen molecules. Dehydroascorbic acid (DHA), the oxidized form of AA, is
Lena Munzel et al.
Autophagy, 17(6), 1458-1478 (2020-06-10)
Coupling of Atg8 to phosphatidylethanolamine is crucial for the expansion of the crescent-shaped phagophore during cargo engulfment. Atg21, a PtdIns3P-binding beta-propeller protein, scaffolds Atg8 and its E3-like complex Atg12-Atg5-Atg16 during lipidation. The crystal structure of Atg21, in complex with the
Lucas B Harrington et al.
Molecular cell, 79(3), 416-424 (2020-07-10)
CRISPR-Cas12c/d proteins share limited homology with Cas12a and Cas9 bacterial CRISPR RNA (crRNA)-guided nucleases used widely for genome editing and DNA detection. However, Cas12c (C2c3)- and Cas12d (CasY)-catalyzed DNA cleavage and genome editing activities have not been directly observed. We
Maria João Correia et al.
Antioxidants (Basel, Switzerland), 10(9) (2021-09-29)
We hypothesized that an interplay between aryl hydrocarbon receptor (AhR) and cysteine-related thiolome at the kidney cortex underlies the mechanisms of (mal)adaptation to chronic intermittent hypoxia (CIH), promoting arterial hypertension (HTN). Using a rat model of CIH-HTN, we investigated the

Articles

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

A RPLC-MS workflow for mass analysis of reduced monoclonal antibody, trastuzumab, including reduction procedures, calibration, and system suitability tests.

See All

Protocols

A complete workflow for the intact and middle-up mass analysis of reduced and non-reduced monoclonal antibodies based on SEC-MS with sample preparation by protein-A affinity clean-up.

A complete workflow for the intact and middle-up mass analysis of reduced and non-reduced monoclonal antibodies based on SEC-MS with sample preparation by protein-A affinity clean-up.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service