Skip to Content
Merck
All Photos(1)

Key Documents

646547

Supelco

Tris(2-carboxyethyl)phosphine hydrochloride solution

0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)

Synonym(s):

TCEP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H15O6P · HCl
CAS Number:
Molecular Weight:
286.65
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.25

Quality Level

concentration

0.5 M

refractive index

n20/D 1.367

pH

7.0(aqueous solution; pH was adjusted with ammonium hydroxide)

density

1.041 g/mL at 25 °C

SMILES string

Cl.OC(=O)CCP(CCC(O)=O)CCC(O)=O

InChI

1S/C9H15O6P.ClH/c10-7(11)1-4-16(5-2-8(12)13)6-3-9(14)15;/h1-6H2,(H,10,11)(H,12,13)(H,14,15);1H

InChI key

PBVAJRFEEOIAGW-UHFFFAOYSA-N

General description

It belongs to the trialkylphosphine class.
Tris (2-carboxyethyl) phosphine (TCEP) is very effective in cleaving disulfide bonds in aqueous solution. It dissolves in water and is odorless, unlike other trialkylphosphines (tributylphosphine). It is also less toxic than 2-mercaptoethanol. These advantages make it better than the other reducing agents.

Application

Tris (2-carboxyethyl) phosphine (TCEP) can be used in several downstream applications including SDS-PAGE, mass spectrometry, labeling with cysteine specific tags, and modification of cysteine containing compounds. It prevents oxidation of protein samples, which makes it a useful buffer component as it helps to preserve enzymatic activity. It has been used in the reduction and measurement of glutathione.
Tris (2-carboxyethyl) phosphine (TCEP) has also been used:
  • to cleave cysteine residues in a synthetic peptide
  • in reduction buffer for RNA Sequential Probing of Targets (SPOTs) imaging
  • for the reduction of oligonucleotides
  • as reducing agent during mitochondrial isolation

Biochem/physiol Actions

As a non-mercaptan reducing agent, it avoids the toxicity inherent in thiol-containing compounds. It is capable of disrupting the botulinum neurotoxin B heavy-chain/light-chain complex that is held together by a single disulfide bond, and that is responsible for endocytosis, and ultimately the toxicity, of the toxin. Since disulfide-coupled subunits are characteristic of many toxins (e.g., ricin, snake venom, and all BoNT serotypes), it may be useful as a rescue prophylactic in cases of toxin administration.
Tris(2-carboxyethyl)phosphine hydrochloride solution reduces the disulfide bonds and leaves other functional groups intact in proteins.

related product

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly multiplexed quantitative mass spectrometry analysis of ubiquitylomes
Rose CM, et al.
Cell Systems, 3(4), 395-403 (2016)
Differential labeling of free and disulfide-bound thiol functions in proteins
Seiwert B, et al.
Journal of the American Society For Mass Spectrometry, 19(1), 1-7 (2008)
E B Getz et al.
Analytical biochemistry, 273(1), 73-80 (1999-08-24)
The newly introduced sulfhydryl reductant tris(2-carboxyethyl)phosphine (TCEP) is a potentially attractive alternative to commonly used dithiothreitol (DTT). We compare properties of DTT and TCEP important in protein biochemistry, using the motor enzyme myosin as an example protein. The reductants equally
Xuerong Shi et al.
Journal of cellular biochemistry, 107(5), 1021-1030 (2009-06-06)
The light chain (LC) of botulinum neurotoxin B (BoNT/B) is unable to enter target neuronal cells by itself. It is brought into the cell in association with the BoNT/B heavy chain (HC) through endocytosis. The BoNT HC-LC subunits are held
Defined DNA/nanoparticle conjugates
Ackerson CJ, et al.
Proceedings of the National Academy of Sciences of the USA, 102(38), 13383-13385 (2005)

Articles

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

Rapid trypsin digest kit yields reliable results in less than 2 hours for mass spectrometry analysis.

See All

Protocols

An optimized LC-MS/MS based workflow for low artifact tryptic digestion and peptide mapping of monoclonal antibody, adalimumab (Humira) using filter assisted sample preparation (FASP).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service