Skip to Content
Merck
All Photos(2)

Key Documents

51799

Supelco

Putrescine

analytical standard

Synonym(s):

1,4-Diaminobutane, 1,4-Butanediamine, Putrescine, Tetramethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)4NH2
CAS Number:
Molecular Weight:
88.15
Beilstein:
605282
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

expl. lim.

9.08 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.457 (lit.)

bp

158-160 °C (lit.)

mp

25-28 °C (lit.)

density

0.877 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

NCCCCN

InChI

1S/C4H12N2/c5-3-1-2-4-6/h1-6H2

InChI key

KIDHWZJUCRJVML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The biosynthesis of spermidine and spermine from putrescine and methionine
Tabor H, et al.
The Journal of Biological Chemistry, 233, 907-914 (1958)
A comparison of the TLC-densitometry and HPLC method for the determination of biogenic amines in fish and fishery products
Shakila J.R, et al.
Food Chemistry, 75, 255-259 (2001)
Advances and Technical Standards in Neurosurgery, Volume 24 (2012)
Barry J Shelp et al.
Plant science : an international journal of experimental plant biology, 193-194, 130-135 (2012-07-17)
4-Aminobutyrate (GABA) accumulates in various plant parts, including bulky fruits such as apples, in response to abiotic stress. It is generally believed that the GABA is derived from glutamate, although a contribution from polyamines is possible. Putrescine, but not spermidine
A Raina et al.
Medical biology, 53(3), 121-147 (1975-06-01)
The biochemistry and biological function of the naturally occurring polyamines, putrescine, spermidine, and spermine, have been reviewed with special reference to animal organisms. These compounds are universally distributed in all living material. Their biosynthesis from ornithine and methionine is accurately

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service