Skip to Content
Merck
All Photos(1)

Key Documents

69602

Sigma-Aldrich

4-Methylumbelliferyl-β-D-glucuronide hydrate

β-glucuronidase substrate, fluorogenic, ≥99.0% (HPLC), powder or crystals, suitable for fluorescence

Synonym(s):

4-Methylumbelliferyl-β-D-glucopyranosiduronic acid, MUG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16O9 · xH2O
CAS Number:
Molecular Weight:
352.29 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Methylumbelliferyl-β-D-glucuronide hydrate, suitable for fluorescence, ≥99.0% (HPLC)

Assay

≥99.0% (HPLC)

form

powder or crystals

impurities

≤0.1% free 4-Methylumbelliferone
≤15% water

solubility

DMSO: 50 mg/mL, clear, colorless to very faintly yellow

fluorescence

λex 365 nm; λem 445 nm in 0.1 M phosphate pH 6.5

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

O.CC1=CC(=O)Oc2cc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(O)=O)ccc12

InChI

1S/C16H16O9.H2O/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;/h2-5,11-14,16,18-20H,1H3,(H,21,22);1H2/t11-,12-,13+,14-,16+;/m0./s1

InChI key

URVSQZMOFUEQAW-YYHOVTOASA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methylumbelliferyl-β-D-glucuronide hydrate (MUG) is a fluorogenic substrate.

Application

4-Methylumbelliferyl-β-D-glucuronide hydrate (MUG) has been used:
  • as a β-glucuronidase (GUS) substrate for fluorimetric analysis of reporter gene expression in Arabidopsis thaliana
  • for the fluorimetric assay for GUS activity
  • to measure liposome lysis by n-octylglucoside

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I George et al.
Canadian journal of microbiology, 47(7), 670-675 (2001-09-08)
Two enzymatic methods based on the measurement of the beta-D-galactosidase activity of total coliforms and the beta-D-glucuronidase activity of Escherichia coli were used to assess coliform levels in wastewater alongside traditional culture-based techniques, which can be biased by the aggregation
R.B. Thompson et al.
Analytical Letters, 18, 1847-1847 (1985)
Nathaniel M Butler et al.
Planta, 236(6), 1747-1755 (2012-08-08)
Polypyrimidine tract-binding (PTB) proteins are RNA-binding proteins that target specific RNAs for post-transcriptional processing by binding cytosine/uracil motifs. PTBs have established functions in a range of RNA processes including splicing, translation, stability and long-distance transport. Six PTB-like genes identified in
Jean-Félix Dallery et al.
Journal of experimental botany, 71(10), 2910-2921 (2020-02-02)
Infection of Arabidopsis thaliana by the ascomycete fungus Colletotrichum higginsianum is characterized by an early symptomless biotrophic phase followed by a destructive necrotrophic phase. The fungal genome contains 77 secondary metabolism-related biosynthetic gene clusters, whose expression during the infection process
Zeng Tao et al.
Nature plants, 5(4), 424-435 (2019-04-10)
Some overwintering plants acquire competence to flower, after experiencing prolonged cold in winter, through a process termed vernalization. In the crucifer plant Arabidopsis thaliana, prolonged cold induces chromatin-mediated silencing of the potent floral repressor FLOWERING LOCUS C (FLC) by Polycomb

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service