Skip to Content
Merck
All Photos(3)

Key Documents

M6018

Sigma-Aldrich

4-Methylumbelliferyl β-D-cellobioside

glucanase substrate, fluorogenic, ≥98.00% (TLC), powder

Synonym(s):

4-Methylumbelliferyl beta-D-cellobioside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28O13
CAS Number:
Molecular Weight:
500.45
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Product Name

4-Methylumbelliferyl β-D-cellobioside, glucanase substrate

Quality Level

Assay

≥98.00% (TLC)

form

powder

solubility

water: 19.60-20.40 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H](CO)[C@@H](O[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H](O)[C@H]3O)ccc12

InChI

1S/C22H28O13/c1-8-4-14(25)32-11-5-9(2-3-10(8)11)31-21-19(30)17(28)20(13(7-24)34-21)35-22-18(29)16(27)15(26)12(6-23)33-22/h2-5,12-13,15-24,26-30H,6-7H2,1H3/t12-,13-,15-,16+,17-,18-,19-,20-,21-,22+/m1/s1

InChI key

PRTGXBPFDYMIJH-MKQZUAMYSA-N

Application

4-Methylumbelliferyl β-D-cellobioside has been used as a substrate:
  • to assay cellulase from A. niger
  • to determine the extracellular enzyme activity for 1,4-β-cellobiohydrolase
  • to assay the activity of bound enzyme, endoglucanase I (EGIc) Trichoderma reesei class II hydrophobins (HFBI)

Biochem/physiol Actions

4-Methylumbelliferyl β-D-cellobioside is used as a substrate for cellulase assays. It is mainly used for activity staining of cellulases in polyacrylamide gels.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G P Xue et al.
Journal of general microbiology, 138(11), 2397-2403 (1992-11-01)
A plant polysaccharide hydrolase cDNA, designated celD, was isolated from a cDNA library of the rumen fungus Neocallimastix patriciarum. The enzyme encoded by celD had endoglucanase, cellobiohydrolase and xylanase activities. Deletion analysis revealed that celD cDNA can be truncated to
Surface adhesion of fusion proteins containing the hydrophobins HFBI and HFBII from Trichoderma reesei
Linder M, et al.
Protein Science, 11(9), 2257-2266 (2002)
A V Gusakov et al.
Biochimica et biophysica acta, 1073(3), 481-485 (1991-04-09)
Using 4-methylumbelliferyl (MUF) beta-D-cellobioside as a substrate, the ability of cellobiohydrolase I from Trichoderma longibrachiatum to catalyze transglycosylation has been demonstrated. At substrate concentrations greater than 2 mM, the formation of MUF-tetrasaccharide was detected using HPLC. In the course of
K H Jung et al.
Biochemistry and molecular biology international, 44(2), 283-292 (1998-04-08)
A Clostridium thermocellum xylanase gene, designated xynX, was cloned in Escherichia coli and was categorized a novel gene as a result of the comparison of restriction patterns of the C. thermocellum xylanase genes so far reported. The xynX gene encodes
M Pedraza-Reyes et al.
Archives of microbiology, 168(4), 321-327 (1997-09-23)
The gram-negative bacterium Myxobacter sp. AL-1 produces chitosanase-cellulase activity that is maximally excreted during the stationary phase of growth. Carboxymethylcellulase zymogram analysis revealed that the enzymatic activity was correlated with two bands of 32 and 35 kDa. Ion-exchange-chromatography-enriched preparations of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service