Skip to Content
Merck
All Photos(1)

Key Documents

63410

Sigma-Aldrich

Malonyl coenzyme A tetralithium salt

≥90% (HPLC)

Synonym(s):

Malonyl-CoA tetralithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H34Li4N7O19P3S
CAS Number:
Molecular Weight:
877.31
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥90% (HPLC)

form

solid

impurities

≤12% water

solubility

H2O: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Li+].[Li+].[Li+].[Li+].CC(C)(COP([O-])(=O)OP([O-])(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1OP([O-])([O-])=O)n2cnc3c(N)ncnc23)[C@@H](O)C(=O)NCCC(=O)NCCSC(=O)CC(O)=O

InChI

1S/C24H38N7O19P3S.4Li/c1-24(2,19(37)22(38)27-4-3-13(32)26-5-6-54-15(35)7-14(33)34)9-47-53(44,45)50-52(42,43)46-8-12-18(49-51(39,40)41)17(36)23(48-12)31-11-30-16-20(25)28-10-29-21(16)31;;;;/h10-12,17-19,23,36-37H,3-9H2,1-2H3,(H,26,32)(H,27,38)(H,33,34)(H,42,43)(H,44,45)(H2,25,28,29)(H2,39,40,41);;;;/q;4*+1/p-4/t12-,17-,18-,19+,23-;;;;/m1..../s1

InChI key

DQLXXYMNINXQSV-QTRRCMKVSA-J

Biochem/physiol Actions

Malonyl Coenzyme A is a Coenzyme A derivative that is utilized in fatty acid and polyketide synthesis and in the transport of α-ketoglutarate across the mitochondrial membrane. Malonyl CoA is formed by the Acetyl CoA Carboxylase-mediated carboxylation of acetyl CoA.

Other Notes

Malonyl-CoA in glucose-stimulated insulin secretion from clonal pancreatic β-cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Corkey et al.
The Journal of biological chemistry, 264(36), 21608-21612 (1989-12-25)
To gain insight into the relationship between acyl coenzyme A (CoA) esters and glucose-induced insulin release, acyl-CoA profiles were determined in clonal pancreatic beta-cells (HIT). A high sensitivity high performance liquid chromatography method was used to measure malonyl, succinyl, beta-hydroxy
Christopher J Arthur et al.
ACS chemical biology, 4(8), 625-636 (2009-06-27)
Malonylation of an acyl carrier protein (ACP) by malonyl Coenzyme A-ACP transacylase (MCAT) is fundamental to bacterial fatty acid biosynthesis. Here, we report the structure of the Steptomyces coelicolor (Sc) fatty acid synthase (FAS) ACP and studies of its binding
Koichiro Kogawa et al.
Journal of plant physiology, 164(7), 886-894 (2006-08-05)
The crude malonyltransferase from the petals of Clitoria ternatea was characterized enzymatically to investigate its role on the biosynthetic pathways of anthocyanins and flavonol glycosides. In C. ternatea, a blue flower cultivars (DB) and mauve flower variety (WM) accumulate polyacylated
Heather Seidle et al.
Journal of bacteriology, 186(8), 2499-2503 (2004-04-03)
Cfa1 was overproduced in Escherichia coli and Pseudomonas syringae, and the degree of 4'-phosphopantetheinylation was determined. The malonyl-coenzyme A:acyl carrier protein transacylase (FabD) of P. syringae was overproduced and shown to catalyze malonylation of Cfa1, suggesting that FabD plays a
Dea Hwan Kim et al.
Plant physiology and biochemistry : PPB, 47(11-12), 991-997 (2009-09-08)
In this study, a flavonoid malonyltransferase (OsMaT-2) was cloned from Oryza sativa, and the recombinant protein OsMaT-2 was purified via affinity chromatography. OsMaT-2 utilized a variety of flavonoid glucosides, including flavanone glucosides, flavone glucosides, flavonol glucosides, and isoflavone glucosides as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service