Skip to Content
Merck
All Photos(3)

Key Documents

62528

Sigma-Aldrich

Lithium hydroxide monohydrate

BioUltra, ≥99.0% (T)

Synonym(s):

LiOH · H2O, Lithium hydroxide, Lithium hydroxide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiOH · H2O
CAS Number:
Molecular Weight:
41.96
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.25
grade:
reagent grade
form:
solid
solubility:
H2O: 1 M at 20 °C, clear, colorless

grade

reagent grade

product line

BioUltra

Assay

≥99.0% (T)

form

solid

impurities

insoluble matter, passes filter test

pH

~12 (25 °C, 1 M in H2O)

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Al: ≤20 mg/kg
As: ≤0.2 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤200 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.02
λ: 280 nm Amax: 0.02

SMILES string

[Li+].O.[OH-]

InChI

1S/Li.2H2O/h;2*1H2/q+1;;/p-1

InChI key

GLXDVVHUTZTUQK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Lithium hydroxide, an alkaline activator/catalyst of synthetic processes, may be used in the formation of polycrystalline Li2O nanowires.

Other Notes

Titrant in the procedure for the cleavage of Asn-Gly bonds with hydroxylamine

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cleavage at Asn-Gly bonds with hydroxylamine.
P Bornstein et al.
Methods in enzymology, 47, 132-145 (1977-01-01)
Jinyoung Jeong et al.
Advanced materials (Deerfield Beach, Fla.), 24(15), 1999-2003 (2012-03-21)
Highly water-soluble and color-tunable photoluminescent fullerene nanoparticles are synthesized by using tetraethylene glycol (TEG) and lithium hydroxide as a catalyst. The maximum PL emission changes depend on the contents of the remaining π-conjugation in oxidized C(60), which is partially covalently
Muhammad Ayoub et al.
Bioresource technology, 112, 308-312 (2012-03-23)
The synthesis of oxygenated fuel additives via solvent freebase-catalyzed etherification of glycerol is reported. The products of glycerol etherification arediglycerol (DG) and triglycerol (TG) with DG being the favorable one. The catalytic activity of different homogeneous alkali catalysts (LiOH, NaOH
Ayorinde Awonusi et al.
Calcified tissue international, 81(1), 46-52 (2007-06-07)
A series of apatites with varying carbonate levels was prepared in order to assign the carbonate bands and calibrate for Raman analysis of natural materials. Overlap of carbonate bands with phosphate peaks was resolved by curve fitting. A peak at
L C Hsu et al.
Journal of hazardous materials, 142(1-2), 242-249 (2006-09-19)
Hexavalent Cr has been identified as one of the toxic metals commonly present in industrial effluents. Among the treatment techniques developed for removing Cr(VI) from waste waters, sorption is most commonly applied, due to its simplicity and efficiency. However, few

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service