Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium hydroxide monohydrate
Linear Formula:
LiOH · H
2
O
CAS Number:
1310-66-3
Molecular Weight:
41.96
EC Number:
215-183-4
Products
Product Category
inorganic bases (2)
salts (2)
Brand
Sigma-Aldrich (6)
Sustainability
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade
ACS reagent (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
402974
Lithium hydroxide monohydrate,
ACS reagent, ≥98.0%
Expand
View Pricing
930903
Lithium hydroxide monohydrate,
battery grade, ≥99.9% trace metals basis
Expand
View Pricing
62528
Lithium hydroxide monohydrate,
BioUltra, ≥99.0% (T)
Expand
View Pricing
450197
Lithium hydroxide monohydrate,
99.995% trace metals basis
Expand
View Pricing
254274
Lithium hydroxide monohydrate,
99.95% trace metals basis
Expand
View Pricing
L4533
Lithium hydroxide monohydrate,
BioXtra, 98.5-101.5% (titration)
Expand
View Pricing