Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetramethylrhodamine isothiocyanate–Dextran
Synonym(s):
TRITC-Dextran
Products
Product Category
carbohydrates (6)
polysaccharides (3)
Brand
Sigma-Aldrich (6)
Biological Source
Color
Physical Form
Application
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
T1037
Tetramethylrhodamine isothiocyanate–Dextran,
average mol wt 4,400
Expand
View Pricing
T1287
Tetramethylrhodamine isothiocyanate–Dextran,
average mol wt 155,000
Expand
View Pricing
T1162
Tetramethylrhodamine isothiocyanate–Dextran,
average mol wt 65,000-85,000
Expand
View Pricing
52194
Tetramethylrhodamine isothiocyanate–Dextran,
average mol wt 500,000
Expand
View Pricing
42874
Tetramethylrhodamine isothiocyanate–Dextran,
average mol wt 40,000
Expand
View Pricing
73766
Tetramethylrhodamine isothiocyanate–Dextran,
average mol wt 20,000
Expand
View Pricing