Skip to Content
Merck
All Photos(1)

Key Documents

50227

Sigma-Aldrich

D-Luciferin potassium salt

≥98.0% (HPLC)

Synonym(s):

(S)-2-(6-Hydroxy-2-benzothiazolyl)-2-thiazoline-4-carboxylic acid potassium salt, 4,5-Dihydro-2-(6-hydroxy-2-benzothiazolyl)-4-thiazolecarboxylic acid potassium salt, Firefly luciferin potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7KN2O3S2
CAS Number:
Molecular Weight:
318.41
Beilstein:
5416262
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98.0% (HPLC)

form

powder or crystals

solubility

water: soluble

storage temp.

−20°C

SMILES string

[K+].Oc1ccc2nc(sc2c1)C3=N[C@H](CS3)C([O-])=O

InChI

1S/C11H8N2O3S2.K/c14-5-1-2-6-8(3-5)18-10(12-6)9-13-7(4-17-9)11(15)16;/h1-3,7,14H,4H2,(H,15,16);/q;+1/p-1/t7-;/m1./s1

InChI key

UMBKGTQQGYPQBE-OGFXRTJISA-M

Looking for similar products? Visit Product Comparison Guide

Application

D-Luciferin may be used as a substrate in luciferase-based (EC 1.13.12.7) bioluminescence imaging and cell-based high-throughput screening applications. ATP can be measured with a reagent made up of D-luciferin and luciferase from firefly.D-Luciferin may be used and studied as a partial agonist for G protein-coupled receptor-35 (GPR35).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A B Madhankumar et al.
PloS one, 13(1), e0181529-e0181529 (2018-01-06)
Peripheral nerve sheath tumors are benign tumors that have the potential to transform into malignant peripheral nerve sheath tumors (MPNSTs). Interleukin-13 receptor alpha 2 (IL13Rα2) is a cancer associated receptor expressed in glioblastoma and other invasive cancers. We analyzed IL13Rα2
Mayu Yasunaga et al.
BMC biotechnology, 17(1), 54-54 (2017-06-24)
Three-dimensional (3D) spheroids are frequently used in toxicological study because their morphology and function closely resemble those of tissue. As these properties are maintained over a long term, repeated treatment of the spheroids with a test object is possible. Generally
Yun-Jung Chiang et al.
Oncogene, 38(9), 1432-1447 (2018-09-30)
High-frequency relapse remains a clinical hurdle for complete remission of T-cell acute lymphoblastic leukemia (T-ALL) patients, with heterogeneous dysregulated signaling profiles-including of Raf-MEK-ERK and Akt-mTORC1-S6K signaling pathways-recently being implicated in disease outcomes. Here we report that GM-CSF/IL-3/IL-5 receptor common β-chain-associated
Jiali Wu et al.
Nature communications, 11(1), 3708-3708 (2020-07-28)
The Cre-loxP recombination system is a powerful tool for genetic manipulation. However, there are widely recognized limitations with chemically inducible Cre-loxP systems, and the UV and blue-light induced systems have phototoxicity and minimal capacity for deep tissue penetration. Here, we
Xinyi Xu et al.
Frontiers in pharmacology, 11, 575-575 (2020-05-21)
Britanin has been reported to have therapeutic effects on neurodegenerative and inflammation-based diseases. However, whether it is involved in the regulation of triple-negative breast cancer development has not been elucidated. In this study, we investigated the anti-tumor activity against triple-negative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service