Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Product Name
Colorimeter Move 100,
technique(s)
photometry: suitable
Quality Level
compatibility
for use with Spectroquant® Move 100
storage temp.
15-25°C
General description
Pre-programmed for over 100 parameters using our high quality Spectroquant® Test Kits this compact and portable colorimeter covers every important parameter for water analysis in one single instrument. This handy, battery-operated instrument comes with a carrying case and is designed for in-process and field work.
Legal Information
Storage Class Code
11 - Combustible Solids
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
Photometric determination using the Griess method subsequent to extraction
Photometric determination with Chromazurole S subsequent to acid mineralisation; DIN decomposition
Method: Photometric determination with 2,2'-dinitro-5,5'-dithiodibenzoic acid (Ellman's reagent)
Photometric determination with Chromazurole S subsequent to acid mineralisation
Related Content
Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.
Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.
Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.
Learn about the sample preparation and photometric determination of non-ionic surfactants in Extran® rinse solutions (MA 01, MA 02, MA 05, AP 22) using Spectroquant® test kits and spectrophotometers.
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service