Skip to Content
Merck
All Photos(1)

Key Documents

808350

Sigma-Aldrich

TokeOni

Synonym(s):

2-[(1E,3E)-4-[4-(Dimethylamino)phenyl]-1,3-butadien-1-yl]-4,5-dihydro-(4S)-4-thiazolecarboxylic acid hydrochloride salt, AkaLumine-HCl, Soluble NIR emission luciferin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2O2S · xHCl
Molecular Weight:
338.85 (free base basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

storage temp.

−20°C

SMILES string

CN(C)C1=CC=C(/C=C/C=C/C2=N[C@@H](C(O)=O)CS2)C=C1.[H]Cl

InChI

1S/C16H18N2O2S.ClH/c1-18(2)13-9-7-12(8-10-13)5-3-4-6-15-17-14(11-21-15)16(19)20;/h3-10,14H,11H2,1-2H3,(H,19,20);1H/b5-3+,6-4+;/t14-;/m1./s1

InChI key

PZCNKVAGZCXXHX-SSRSOBHISA-N

General description

  • Luciferin is a generic term for the light-emitting compound found in organisms that generate bioluminescence.
  • TokeOni is a new NIR emission luciferin analogue having luminescence peak at 670 ~ 680 nm with a peak range in the near-infrared (NIR) region.
  • TokeOni has the enhanced solubility of >10 mg/mL compared to HCl free (0.2 mg/mL).

Application

  • Bioimaging.
  • in vivo imaging.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of simple firefly luciferin analogs emitting blue, green, red and near-infrared biological window light.
Satoshi I, et al.
Tetrahedron, 69(19), 3847-3856 (2013)
Synthesis and luminescence properties of biphenyl-type firefly luciferin analogs with a new, near-infrared light-emitting bioluminophore.
Chihiro M, et al.
Tetrahedron, 69(46), 9726-9734 (2013)
Gi Bum Kim et al.
Cell, 179(1), 251-267 (2019-09-21)
In situ transgenesis methods such as viruses and electroporation can rapidly create somatic transgenic mice but lack control over copy number, zygosity, and locus specificity. Here we establish mosaic analysis by dual recombinase-mediated cassette exchange (MADR), which permits stable labeling

Articles

Bioluminescence imaging (BLI) systems allows for high-sensitive and noninvasive monitoring of cell proliferation, activity of signaling pathways and protein-protein interactions in living tissues.

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

seMpai substrate is suitable for near-infrared BLI in biological experiments, offering high solubility for extended bioluminescent applications.

See All

Related Content

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Organic electronics utilizes organic conductors and semiconductors for applications in organic photovoltaics, organic light-emitting diodes, and organic field-effect transistors.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service