Skip to Content
Merck
All Photos(3)

Key Documents

49767

Sigma-Aldrich

Glycerol

BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)

Synonym(s):

1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
50161901
PubChem Substance ID:
NACRES:
NA.25
grade:
Molecular Biology
for molecular biology
technique(s):
GC/GC: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)

grade

Molecular Biology
for molecular biology

Quality Level

vapor density

3.1 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

BioUltra

Assay

≥99.5% (GC)

form

viscous liquid

autoignition temp.

698 °F

technique(s)

GC/GC: suitable

impurities

DNases, none detected
RNases, none detected
insoluble matter, passes filter test
phosphatases, none detected
proteases, none detected

ign. residue

≤0.1%

refractive index

n20/D 1.474 (lit.)
n20/D 1.474

pH

5.5-8 (25 °C, 5 M in H2O)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

solubility

H2O: 5 M at 20 °C, clear, colorless

density

1.25 g/mL (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
sulfate (SO42-): ≤10 mg/kg

cation traces

Ag: ≤5 mg/kg
Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
NH4+: ≤5 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Tl: ≤5 mg/kg
Zn: ≤1 mg/kg

SMILES string

OCC(O)CO

λ

5 M in H2O

UV absorption

λ: 260 nm Amax: 0.05
λ: 280 nm Amax: 0.07

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glycerol is odourless, colorless, viscous in nature, and exists as a sweet tasting liquid. It can be derived naturally as well as from petrochemical feedstock. Glycerol has a wide variety of applications, and is one of the most valuable and versatile chemical substances in nature. It can be used as an emollient, solvent, sweetening agent, in pharmaceutical formulations, cosmetics, foodstuffs and toiletries. It is very stable and can be easily stored under normal temperature; also it is non-irritating and has no adverse impact to the environment.

Application

Glycerol is used both in sample preparation and gel formation for polyacrylamide gel electrophoresis. Glycerol (5-10%) increases the density of a sample so that the sample will layer at the bottom of a gel′s sample well. Glycerol is also used to aid in casting gradient gels and as a protein stabilizer and storage buffer component.

Biochem/physiol Actions

Glycerol has been used as
  • a supplement during cell culture of Mycobacterium tuberculosis and Mycobacterium avium.
  • a fuel during designing enzymatic biofuel cell.
  • a liquid composite matrix with 4-HCCA and 3-amino­quinoline for analysis of neutral and acidic glycans.
  • a matrix for fast atom bombardment MS.
  • may be employed as liquid matrix for the quantification studies by MALDI (Matrix-assisted laser desorption/ionization mass spectrometry) analysis.
Glycerol is hygroscopic in nature and is soluble in water owing to its three hydrophilic alcoholic hydroxyl groups. It can form both inter- and intramolecular hydrogen bonds, making it a very flexible molecule. The physiologic effect of glycerine is due to cell-mediated immunity, increased IgG production and increased histamine release.

Other Notes

Component of loading buffer in agarose gel electrophoresis of nucleic acids previously denatured with glyoxal; Preparation of phage and plasmid DNA, for the storage of pure cultures

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R.C. Ogden et al.
Methods in Enzymology, 152, 79-79 (1987)
Practical aspects of preparing phage and plasmid DNA: growth, maintenance, and storage of bacteria and bacteriophage.
H Miller
Methods in enzymology, 152, 145-170 (1987-01-01)
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Benoît Ranchoux et al.
The American journal of pathology, 185(2), 356-371 (2014-12-17)
Pulmonary veno-occlusive disease (PVOD) is an uncommon form of pulmonary hypertension (PH) characterized by progressive obstruction of small pulmonary veins and a dismal prognosis. Limited case series have reported a possible association between different chemotherapeutic agents and PVOD. We evaluated
T W Chew et al.
Oncogene, 33(21), 2717-2727 (2013-07-03)
RAS and Rho small GTPases are key molecular switches that control cell dynamics, cell growth and tissue development through their distinct signaling pathways. Although much has been learnt about their individual functions in both cell and animal models, the physiological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service