Skip to Content
Merck
All Photos(1)

Key Documents

36791

Supelco

Chlorothalonil

PESTANAL®, analytical standard

Synonym(s):

Tetrachloroisophthalodinitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8Cl4N2
CAS Number:
Molecular Weight:
265.91
Beilstein:
1978326
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Clc1c(Cl)c(C#N)c(Cl)c(C#N)c1Cl

InChI

1S/C8Cl4N2/c9-5-3(1-13)6(10)8(12)7(11)4(5)2-14

InChI key

CRQQGFGUEAVUIL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlorothalonil is a an organochlorine, broad-spectrum and non-systemic fungicide, which can be widely used in agriculture, in order to have a control on foliar pathogens.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of chlorothalonil photodegradation in natural waters and in the presence of humic substances
Sakkas.AV, et al.
Chemosphere, 48, 939-945 (2002)
Transformation pathways of 14C-chlorothalonil in tropical soils
Regitano.B.J, et al.
Archives of Environmental Contamination and Toxicology, 40, 295-302 (2001)
Xing-Hai Liu et al.
European journal of medicinal chemistry, 44(7), 2782-2786 (2009-02-28)
A series of cyclopropanecarboxamide were prepared and tested for antifungal activity in vivo. The preliminary bioassays indicated that some compounds are comparable to the commercial fungicides. To further explore the comprehensive structure-activity relationship on the basis of fungicidal activity data
Zining Cui et al.
Bioorganic & medicinal chemistry letters, 21(23), 7193-7196 (2011-10-19)
Assuming that the water solubility of our previous hydrazone derivatives would improve after modification with sugars while keeping or modulating their notable biological activities, we designed and synthesized some glycosyl hydrazine and hydrazone derivatives. Bioassay results indicated that the antitumor
Xiangwei Wu et al.
Ecotoxicology and environmental safety, 81, 65-69 (2012-05-11)
The effect of successive chlorothalonil applications on the persistence of chlorothalonil, soil respiration activity, and dehydrogenase activity (DHA) in soil was investigated under laboratory conditions. The persistence of chlorothalonil in soil was prolonged significantly with the increase in the concentration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service