Skip to Content
Merck
All Photos(1)

Key Documents

45708

Supelco

Ziram

PESTANAL®, analytical standard

Synonym(s):

Zinc dimethyldithiocarbamate, Dimethyldithiocarbamic acid zinc salt, Ziram

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCSSZnSCSN(CH3)2
CAS Number:
Molecular Weight:
305.82
Beilstein:
3707008
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

reaction suitability

reagent type: catalyst
core: zinc

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

248-257 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CN(C)C(=S)S[Zn]SC(=S)N(C)C

InChI

1S/2C3H7NS2.Zn/c2*1-4(2)3(5)6;/h2*1-2H3,(H,5,6);/q;;+2/p-2

InChI key

DUBNHZYBDBBJHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Ziram is a fungicide, widely used for the protection of vegetable crops, seed foliage and fruits. It is also employed in rubber industry as an antioxidant and vulcanization accelerator.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ziram may be used as an analytical reference standard for the quantification of the analyte in environmental samples, grains, and vegetable samples using different analytical techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Liver,Blood, Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of ziram in vegetable samples by square wave voltammetry
Qui P, et al.
Chinese Chemical Letters = Zhongguo Hua Xue Kuai Bao, 19(11), 1337-1340 (2008)
Spectrophotometric determination of ziram (dithiocarbamate fungicide) by thiocyanate and rhodamine 6G method
Mathew L, et al.
Talanta, 42(1), 41-43 (1995)
Josiane P Lafleur et al.
Lab on a chip, 12(22), 4651-4656 (2012-07-25)
Conventional methods of environmental analysis can be significantly improved by the development of portable microscale technologies for direct in-field sensing at remote locations. This report demonstrates the vast potential of gold nanoparticle-based microfluidic sensors for the rapid, in-field, detection of
Thyneice R Taylor et al.
Toxicology mechanisms and methods, 21(8), 577-584 (2011-08-24)
Human natural killer (NK) cells are central in immune defense with their ability to lyse tumor cells and virally infected cells. Tumor formation and viral infection may increase if NK cytotoxic function is disrupted. Ziram (zinc dithiocarbamate) is used as
Gha-Young Kim et al.
Journal of environmental monitoring : JEM, 10(5), 632-637 (2008-05-02)
A highly sensitive enzyme electrode was prepared based on gold nanoparticles for measurement of pesticides. Gold nanoparticles of 25-30 nm were synthesized on a glassy carbon electrode by double-pulse technique while the coverage was controlled by applied potential and time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service